Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135050.1 | NP_001128522.1 | 3 - 4 | 448 | 73 |
NM_020789.3 | NP_065840.2 | 3 - 4 | 448 | 73 | |
XM_017001884.1 | XP_016857373.1 | 3 - 4 | 513 | 73 | |
GenBank mRNA | AB037776.1 | - | 3 - 4 | 448 | 73 |
AY358128.1 | - | 3 - 4 | 466 | 73 | |
BC030141.2 | - | 3 - 4 | 448 | 73 |