Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142725.1 | NP_001136197.1 | 9 - 9 | 1804 | 122 |
GenBank mRNA | AB040937.1 | - | 1 - 1 | 2151 | 122 |
AK127391.1 | - | 9 - 9 | 951 | 122 | |
AK292594.1 | - | 10 - 10 | 2017 | 122 | |
AK304347.1 | - | 8 - 8 | 1724 | 122 |