Assay Design
Gene Transcripts
- Gene Symbol
- LRRC4C
- Entrez Gene ID
-
57689
- Gene Name
- leucine rich repeat containing 4C
- Gene Aliases
-
NGL-1, NGL1
- Chromosome Location
-
Chr.11: 40107066 - 41460212 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_047674.1 |
|
1 - 2 |
945 |
109 |
GenBank mRNA |
AB046800.1 |
- |
1 - 2 |
1930 |
109 |
AY358293.1 |
- |
1 - 2 |
776 |
109 |
View Full Product Details