Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128933.2 | NP_001122405.1 | 3 - 4 | 1267 | 102 |
NM_001128934.2 | NP_001122406.1 | 3 - 4 | 1267 | 102 | |
NM_001286754.1 | NP_001273683.1 | 3 - 4 | 1219 | 102 | |
NM_133477.2 | NP_597734.2 | 3 - 4 | 1267 | 102 | |
GenBank mRNA | AJ010482.1 | - | 1 - 2 | 287 | 102 |
AK307711.1 | - | 3 - 4 | 1263 | 102 | |
AL832031.1 | - | 3 - 4 | 1250 | 102 | |
AL832363.1 | - | 3 - 4 | 1161 | 102 | |
AL833294.1 | - | 3 - 4 | 1253 | 102 | |
AL833547.1 | - | 3 - 4 | 1267 | 102 | |
BC136256.1 | - | 3 - 4 | 1142 | 102 | |
BC150629.1 | - | 3 - 4 | 1266 | 102 | |
BX647849.1 | - | 1 - 2 | 776 | 102 | |
CR936673.1 | - | 3 - 4 | 1259 | 102 | |
EU481975.1 | - | 3 - 4 | 1071 | 102 | |
FN422000.1 | - | 1 - 2 | 322 | 102 | |
KF147165.1 | - | 3 - 4 | 1219 | 102 |