Assay Design



Gene Transcripts

Gene Symbol
CCDC40
Entrez Gene ID
55036
Gene Name
coiled-coil domain containing 40
Gene Aliases
CILD15, FAP172
Chromosome Location
Chr.17: 80036632 - 80100613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017950.3 NP_060420.2 17 - 18 2862 87
XM_011524963.2 XP_011523265.1 17 - 18 2899 87
XM_011524964.2 XP_011523266.1 11 - 12 1831 87
GenBank mRNA AB046860.1 - 13 - 14 2368 87
AK056583.1 - 6 - 7 1241 87
AL713758.1 - 4 - 5 3082 87

View Full Product Details