Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | 11 | 1817 | NP_001107849.1 |
NM_014009.3 | 12 | 1922 | NP_054728.2 | |
XM_006724533.2 | 11 | 2097 | XP_006724596.2 | |
XM_011543916.2 | 11 | 3527 | XP_011542218.1 | |
XM_017029565.1 | 13 | 3383 | XP_016885054.1 | |
XM_017029566.1 | 12 | 3347 | XP_016885055.1 | |
XM_017029567.1 | 10 | 1922 | XP_016885056.1 | |
AF277993.1 | AAG53607.1 | |||
BQ184335.1 | ||||
DB342786.1 | 1 | 458 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | ChrX:49205142 - 49320261 on Build GRCh38 | Gain | CACNA1F PPP1R3F FOXP3 CCDC22 GAGE10 |
nsv528292 | ChrX:48433286 - 49274368 on Build GRCh38 | Gain | PPP1R3F FOXP3 CCDC22 KCND1 CCDC120 WDR45 PRAF2 ERAS SLC38A5 LOC101927635 PQBP1 EBP WAS TFE3 WDR13 SYP PLP2 SUV39H1 FTSJ1 HDAC6 SYP-AS1 SLC35A2 GATA1 PORCN TIMM17B PCSK1N GLOD5 GPKOW CACNA1F MAGIX GRIPAP1 RBM3 PRICKLE3 OTUD5 PIM2 TBC1D25 |
esv32909 | ChrX:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SPACA5 PPP1R3F FOXP3 CCDC22 KCND1 CCDC120 WDR45 PRAF2 ERAS SLC38A5 SSX5 LOC101927635 PQBP1 EBP SSX9 WAS TFE3 WDR13 SPACA5B ZNF182 SYP PLP2 SUV39H1 ZNF630 SSX3 SSX4B FTSJ1 SSX1 HDAC6 SYP-AS1 SLC35A2 SSX6 GATA1 PORCN TIMM17B PCSK1N GLOD5 GPKOW CACNA1F MAGIX GRIPAP1 ZNF630-AS1 RBM3 SSX4 PRICKLE3 OTUD5 PIM2 TBC1D25 |
nsv469734 | ChrX:49205141 - 49320261 on Build GRCh38 | Gain+Loss | CACNA1F PPP1R3F FOXP3 CCDC22 GAGE10 |
nsv6895 | ChrX:49227319 - 49262690 on Build GRCh38 | Insertion | CACNA1F FOXP3 CCDC22 |