Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46617556-46618057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1 XM_011520028.2

Gene Symbol
HARBI1
Entrez Gene ID
283254
Gene Name
harbinger transposase derived 1
Gene Aliases
C11orf77
UniGene
Hs.714463
Gene Chromosome Location
Chr.11: 46603027-46617651 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1 XM_011520028.2

SNP Detail

SNP ID Chromosome Location
rs942703613 Chr.11: 46617700-46617700 on Build GRCh38
rs574379493 Chr.11: 46617736-46617736 on Build GRCh38
rs986944969 Chr.11: 46617961-46617961 on Build GRCh38
rs576682604 Chr.11: 46617632-46617632 on Build GRCh38
rs919202027 Chr.11: 46617650-46617650 on Build GRCh38
rs959569616 Chr.11: 46617893-46617893 on Build GRCh38
rs984746941 Chr.11: 46618001-46618001 on Build GRCh38
rs73451811 Chr.11: 46617973-46617973 on Build GRCh38
rs950507956 Chr.11: 46617581-46617581 on Build GRCh38
rs935432242 Chr.11: 46617648-46617648 on Build GRCh38
rs990465972 Chr.11: 46617904-46617904 on Build GRCh38
rs540059893 Chr.11: 46617603-46617603 on Build GRCh38
rs1007646511 Chr.11: 46617750-46617750 on Build GRCh38
rs545619141 Chr.11: 46617874-46617874 on Build GRCh38
rs983580939 Chr.11: 46617595-46617595 on Build GRCh38
rs971440110 Chr.11: 46617936-46617936 on Build GRCh38
rs1039958463 Chr.11: 46617716-46617716 on Build GRCh38
rs556773501 Chr.11: 46617631-46617631 on Build GRCh38
rs1040832920 Chr.11: 46618020-46618020 on Build GRCh38
rs888988348 Chr.11: 46617744-46617744 on Build GRCh38
rs868574052 Chr.11: 46617636-46617636 on Build GRCh38
rs923806187 Chr.11: 46617930-46617930 on Build GRCh38
rs935153467 Chr.11: 46617940-46617940 on Build GRCh38
rs1009584182 Chr.11: 46617766-46617766 on Build GRCh38
rs1034759887 Chr.11: 46617873-46617873 on Build GRCh38
rs544341630 Chr.11: 46617694-46617694 on Build GRCh38
rs539847018 Chr.11: 46617761-46617761 on Build GRCh38
rs771004353 Chr.11: 46618009-46618009 on Build GRCh38
rs1039693733 Chr.11: 46617992-46617992 on Build GRCh38
rs568414286 Chr.11: 46617983-46617983 on Build GRCh38
rs958455639 Chr.11: 46617620-46617620 on Build GRCh38
rs968331330 Chr.11: 46617820-46617820 on Build GRCh38
rs946554628 Chr.11: 46617639-46617639 on Build GRCh38
rs28365974 Chr.11: 46617696-46617696 on Build GRCh38
rs532286710 Chr.11: 46617857-46617857 on Build GRCh38
rs527731103 Chr.11: 46617997-46617997 on Build GRCh38
rs367698316 Chr.11: 46617740-46617740 on Build GRCh38
rs930541355 Chr.11: 46617975-46617975 on Build GRCh38
rs916498550 Chr.11: 46617638-46617638 on Build GRCh38
rs560440788 Chr.11: 46617767-46617767 on Build GRCh38
rs547465906 Chr.11: 46618010-46618010 on Build GRCh38
rs919901309 Chr.11: 46618025-46618025 on Build GRCh38
rs577965977 Chr.11: 46617974-46617974 on Build GRCh38
rs1000952883 Chr.11: 46617861-46617861 on Build GRCh38
rs923746745 Chr.11: 46617641-46617641 on Build GRCh38
rs1021600369 Chr.11: 46617780-46617780 on Build GRCh38
rs61884264 Chr.11: 46618032-46618032 on Build GRCh38
rs557923082 Chr.11: 46617908-46617908 on Build GRCh38
rs1004641708 Chr.11: 46617846-46617846 on Build GRCh38
rs1040506366 Chr.11: 46617758-46617758 on Build GRCh38
rs979274865 Chr.11: 46617640-46617640 on Build GRCh38
rs887233779 Chr.11: 46617829-46617829 on Build GRCh38
rs968012110 Chr.11: 46617914-46617914 on Build GRCh38
rs748060417 Chr.11: 46617989-46617989 on Build GRCh38
rs960399664 Chr.11: 46617885-46617885 on Build GRCh38
rs183754521 Chr.11: 46617598-46617598 on Build GRCh38
rs746865621 Chr.11: 46617901-46617901 on Build GRCh38
rs562361697 Chr.11: 46617898-46617898 on Build GRCh38
rs531367730 Chr.11: 46617922-46617922 on Build GRCh38
rs781241429 Chr.11: 46617927-46617927 on Build GRCh38

View Full Product Details