Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47468188-47468688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs578053892 Chr.11: 47468561-47468561 on Build GRCh38
rs113708740 Chr.11: 47468480-47468480 on Build GRCh38
rs897982287 Chr.11: 47468549-47468549 on Build GRCh38
rs1056144610 Chr.11: 47468524-47468524 on Build GRCh38
rs763312224 Chr.11: 47468423-47468423 on Build GRCh38
rs950297284 Chr.11: 47468532-47468532 on Build GRCh38
rs896150167 Chr.11: 47468558-47468558 on Build GRCh38
rs749396265 Chr.11: 47468482-47468482 on Build GRCh38
rs912531669 Chr.11: 47468355-47468355 on Build GRCh38
rs967232283 Chr.11: 47468455-47468455 on Build GRCh38
rs940418502 Chr.11: 47468518-47468518 on Build GRCh38
rs561549938 Chr.11: 47468300-47468300 on Build GRCh38
rs543768595 Chr.11: 47468651-47468651 on Build GRCh38
rs1023846662 Chr.11: 47468229-47468229 on Build GRCh38
rs538714231 Chr.11: 47468481-47468481 on Build GRCh38
rs370403413 Chr.11: 47468297-47468297 on Build GRCh38
rs551554566 Chr.11: 47468618-47468618 on Build GRCh38
rs953383058 Chr.11: 47468323-47468326 on Build GRCh38
rs531234987 Chr.11: 47468233-47468233 on Build GRCh38
rs1000984552 Chr.11: 47468264-47468264 on Build GRCh38
rs528422329 Chr.11: 47468278-47468279 on Build GRCh38
rs566992839 Chr.11: 47468398-47468398 on Build GRCh38
rs986778487 Chr.11: 47468316-47468316 on Build GRCh38
rs1036488890 Chr.11: 47468261-47468262 on Build GRCh38
rs965938985 Chr.11: 47468377-47468377 on Build GRCh38
rs536966048 Chr.11: 47468641-47468641 on Build GRCh38
rs879679788 Chr.11: 47468213-47468218 on Build GRCh38
rs1055873653 Chr.11: 47468483-47468483 on Build GRCh38
rs968612373 Chr.11: 47468251-47468251 on Build GRCh38
rs994985161 Chr.11: 47468587-47468587 on Build GRCh38
rs571078440 Chr.11: 47468624-47468624 on Build GRCh38
rs199995378 Chr.11: 47468227-47468228 on Build GRCh38
rs770272288 Chr.11: 47468440-47468440 on Build GRCh38
rs369566397 Chr.11: 47468230-47468231 on Build GRCh38
rs377227389 Chr.11: 47468298-47468298 on Build GRCh38
rs142163892 Chr.11: 47468392-47468392 on Build GRCh38
rs746736658 Chr.11: 47468343-47468343 on Build GRCh38
rs1045327061 Chr.11: 47468582-47468582 on Build GRCh38
rs1036603418 Chr.11: 47468546-47468546 on Build GRCh38
rs774484632 Chr.11: 47468609-47468609 on Build GRCh38
rs1028178766 Chr.11: 47468304-47468304 on Build GRCh38
rs1031482685 Chr.11: 47468265-47468265 on Build GRCh38
rs115779652 Chr.11: 47468379-47468379 on Build GRCh38

View Full Product Details