Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47468526-47469069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs897982287 Chr.11: 47468549-47468549 on Build GRCh38
rs896150167 Chr.11: 47468558-47468558 on Build GRCh38
rs764640041 Chr.11: 47468732-47468732 on Build GRCh38
rs3087947 Chr.11: 47468714-47468714 on Build GRCh38
rs747293921 Chr.11: 47468896-47468899 on Build GRCh38
rs552684504 Chr.11: 47468779-47468779 on Build GRCh38
rs1049698804 Chr.11: 47468948-47468948 on Build GRCh38
rs974660885 Chr.11: 47468761-47468761 on Build GRCh38
rs543768595 Chr.11: 47468651-47468651 on Build GRCh38
rs551554566 Chr.11: 47468618-47468618 on Build GRCh38
rs917616175 Chr.11: 47468852-47468852 on Build GRCh38
rs775835336 Chr.11: 47468693-47468693 on Build GRCh38
rs754335211 Chr.11: 47468880-47468880 on Build GRCh38
rs957016174 Chr.11: 47468694-47468694 on Build GRCh38
rs146701169 Chr.11: 47468895-47468896 on Build GRCh38
rs572904928 Chr.11: 47468835-47468835 on Build GRCh38
rs536966048 Chr.11: 47468641-47468641 on Build GRCh38
rs964006575 Chr.11: 47468734-47468734 on Build GRCh38
rs994985161 Chr.11: 47468587-47468587 on Build GRCh38
rs936905937 Chr.11: 47468938-47468938 on Build GRCh38
rs967178497 Chr.11: 47468715-47468715 on Build GRCh38
rs561614150 Chr.11: 47469014-47469014 on Build GRCh38
rs148269523 Chr.11: 47468924-47468924 on Build GRCh38
rs1045327061 Chr.11: 47468582-47468582 on Build GRCh38
rs889329461 Chr.11: 47468956-47468956 on Build GRCh38
rs190722264 Chr.11: 47468871-47468871 on Build GRCh38
rs578053892 Chr.11: 47468561-47468561 on Build GRCh38
rs999991162 Chr.11: 47469043-47469043 on Build GRCh38
rs1019645524 Chr.11: 47469028-47469028 on Build GRCh38
rs943440340 Chr.11: 47468959-47468960 on Build GRCh38
rs767684730 Chr.11: 47468686-47468686 on Build GRCh38
rs901825761 Chr.11: 47468977-47468977 on Build GRCh38
rs35996141 Chr.11: 47468723-47468723 on Build GRCh38
rs141316553 Chr.11: 47468958-47468958 on Build GRCh38
rs752058606 Chr.11: 47468714-47468715 on Build GRCh38
rs966067234 Chr.11: 47468692-47468692 on Build GRCh38
rs1044726759 Chr.11: 47468882-47468882 on Build GRCh38
rs947680971 Chr.11: 47468878-47468878 on Build GRCh38
rs951968315 Chr.11: 47468775-47468775 on Build GRCh38
rs1040490578 Chr.11: 47468959-47468962 on Build GRCh38
rs571078440 Chr.11: 47468624-47468624 on Build GRCh38
rs907659945 Chr.11: 47468798-47468798 on Build GRCh38
rs187754172 Chr.11: 47468692-47468692 on Build GRCh38
rs1009587894 Chr.11: 47469009-47469009 on Build GRCh38
rs949747762 Chr.11: 47468874-47468874 on Build GRCh38
rs958700120 Chr.11: 47468832-47468832 on Build GRCh38
rs146213933 Chr.11: 47468883-47468883 on Build GRCh38
rs1056217112 Chr.11: 47468844-47468844 on Build GRCh38
rs892633751 Chr.11: 47469012-47469012 on Build GRCh38
rs921825942 Chr.11: 47468763-47468763 on Build GRCh38
rs774484632 Chr.11: 47468609-47468609 on Build GRCh38
rs919523798 Chr.11: 47468907-47468907 on Build GRCh38
rs563650592 Chr.11: 47468724-47468724 on Build GRCh38
rs972266899 Chr.11: 47468888-47468888 on Build GRCh38

View Full Product Details