Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47470937-47471442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs1038011815 Chr.11: 47471307-47471307 on Build GRCh38
rs923584163 Chr.11: 47470995-47470995 on Build GRCh38
rs3508 Chr.11: 47471379-47471379 on Build GRCh38
rs1049931891 Chr.11: 47471291-47471291 on Build GRCh38
rs758429691 Chr.11: 47471231-47471232 on Build GRCh38
rs963625184 Chr.11: 47471339-47471339 on Build GRCh38
rs1024446492 Chr.11: 47471005-47471005 on Build GRCh38
rs980174527 Chr.11: 47471096-47471096 on Build GRCh38
rs915154291 Chr.11: 47471181-47471181 on Build GRCh38
rs532394202 Chr.11: 47471236-47471236 on Build GRCh38
rs151250368 Chr.11: 47471389-47471389 on Build GRCh38
rs552203247 Chr.11: 47471290-47471290 on Build GRCh38
rs889486550 Chr.11: 47471301-47471301 on Build GRCh38
rs906743160 Chr.11: 47471160-47471160 on Build GRCh38
rs576886306 Chr.11: 47470989-47470989 on Build GRCh38
rs1005650786 Chr.11: 47471304-47471304 on Build GRCh38
rs566693824 Chr.11: 47471206-47471206 on Build GRCh38
rs971474939 Chr.11: 47471075-47471075 on Build GRCh38
rs974692758 Chr.11: 47471357-47471357 on Build GRCh38
rs1012377115 Chr.11: 47471131-47471131 on Build GRCh38
rs935009474 Chr.11: 47471030-47471030 on Build GRCh38
rs770393256 Chr.11: 47471267-47471267 on Build GRCh38
rs897419281 Chr.11: 47471329-47471329 on Build GRCh38
rs1034866402 Chr.11: 47471410-47471410 on Build GRCh38
rs921750229 Chr.11: 47471371-47471371 on Build GRCh38
rs775169126 Chr.11: 47470997-47470997 on Build GRCh38
rs894057517 Chr.11: 47471127-47471127 on Build GRCh38
rs142431972 Chr.11: 47471373-47471373 on Build GRCh38
rs537953599 Chr.11: 47471341-47471341 on Build GRCh38
rs777061615 Chr.11: 47471040-47471040 on Build GRCh38
rs957583084 Chr.11: 47471152-47471152 on Build GRCh38
rs1045205972 Chr.11: 47471140-47471140 on Build GRCh38
rs945258459 Chr.11: 47471207-47471207 on Build GRCh38
rs568839507 Chr.11: 47471302-47471302 on Build GRCh38
rs927344494 Chr.11: 47471148-47471148 on Build GRCh38
rs148803083 Chr.11: 47470973-47470973 on Build GRCh38
rs994383986 Chr.11: 47471336-47471336 on Build GRCh38
rs546769116 Chr.11: 47471114-47471114 on Build GRCh38
rs765966084 Chr.11: 47471391-47471391 on Build GRCh38
rs1042203785 Chr.11: 47471214-47471214 on Build GRCh38
rs34258212 Chr.11: 47471311-47471312 on Build GRCh38
rs990143235 Chr.11: 47471165-47471165 on Build GRCh38

View Full Product Details