Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080378.1 | NP_001073847.1 | 1 - 2 | 147 | 60 |
NM_152410.2 | NP_689623.2 | 1 - 2 | 147 | 60 | |
XM_017010275.1 | XP_016865764.1 | 1 - 2 | 147 | 60 | |
GenBank mRNA | AK057286.1 | - | 1 - 2 | 147 | 60 |
BC044227.1 | - | 1 - 2 | 134 | 60 |