Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002266.2 | NP_001002266.1 | 3 - 4 | 414 | 104 |
NM_001282866.1 | NP_001269795.1 | 3 - 4 | 414 | 104 | |
NM_145021.5 | NP_659458.2 | 3 - 4 | 902 | 104 | |
XM_005271804.2 | XP_005271861.1 | 3 - 4 | 903 | 104 | |
XM_006717704.2 | XP_006717767.1 | 3 - 4 | 318 | 104 | |
XM_011539492.2 | XP_011537794.1 | 4 - 5 | 954 | 104 | |
XM_011539493.2 | XP_011537795.1 | 5 - 6 | 1195 | 104 | |
XM_011539494.2 | XP_011537796.1 | 4 - 5 | 1144 | 104 | |
XM_011539495.1 | XP_011537797.1 | 3 - 4 | 298 | 104 | |
XM_017015894.1 | XP_016871383.1 | 4 - 5 | 954 | 104 | |
GenBank mRNA | AK057289.1 | - | 2 - 3 | 73 | 104 |
AK307667.1 | - | 3 - 4 | 414 | 104 | |
AK313340.1 | - | 3 - 4 | 371 | 104 | |
BC025394.2 | - | 3 - 4 | 391 | 104 | |
BC066988.1 | - | 3 - 4 | 903 | 104 | |
BG717635.1 | - | 3 - 4 | 396 | 104 | |
BM906478.1 | - | - | 584 | 104 |