Assay Design
Gene Transcripts
- Gene Symbol
- STIM1
- Entrez Gene ID
-
6786
- Gene Name
- stromal interaction molecule 1
- Gene Aliases
-
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
- Chromosome Location
-
Chr.11: 3855703 - 4093210 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK057599.1 |
- |
1 - 1 |
56 |
75 |
View Full Product Details