Assay Design



Gene Transcripts

Gene Symbol
U2SURP
Entrez Gene ID
23350
Gene Name
U2 snRNP associated SURP domain containing
Gene Aliases
SR140, fSAPa
Chromosome Location
Chr.3: 143001524 - 143060725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080415.1 NP_001073884.1 16 - 17 1711 94
NM_001320219.1 NP_001307148.1 16 - 17 1708 94
NM_001320220.1 NP_001307149.1 15 - 16 1667 94
NM_001320222.1 NP_001307151.1 17 - 18 1776 94
XM_005247248.4 XP_005247305.1 16 - 17 1717 94
XM_017006037.1 XP_016861526.1 12 - 13 1319 94
XM_017006038.1 XP_016861527.1 15 - 16 1603 94
XM_017006039.1 XP_016861528.1 9 - 10 1046 94
GenBank mRNA AB002330.1 - 16 - 17 1610 94
AK057679.1 - 3 - 4 1705 94
AK074327.1 - 9 - 10 975 94
BC006474.1 - 16 - 17 1607 94
BC111692.1 - 15 - 16 2038 94

View Full Product Details