Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198625.1 | NP_001185554.1 | 2 - 2 | 408 | 75 |
NM_001198626.1 | NP_001185555.1 | 2 - 2 | 408 | 75 | |
NM_001198627.1 | NP_001185556.1 | 2 - 2 | 408 | 75 | |
NM_001198629.1 | NP_001185558.1 | 1 - 1 | 191 | 75 | |
NM_001198630.1 | NP_001185559.1 | 1 - 1 | 191 | 75 | |
NM_001198632.1 | NP_001185561.1 | 1 - 1 | 191 | 75 | |
NM_001198679.1 | NP_001185608.1 | 2 - 2 | 408 | 75 | |
NM_175635.2 | NP_783553.1 | 1 - 1 | 191 | 75 | |
XM_017013930.1 | XP_016869419.1 | 2 - 2 | 609 | 75 | |
XM_017013932.1 | XP_016869421.1 | 1 - 1 | 159 | 75 | |
XM_017013933.1 | XP_016869422.1 | 1 - 1 | 163 | 75 | |
XM_017013934.1 | XP_016869423.1 | 2 - 2 | 367 | 75 | |
XM_017013935.1 | XP_016869424.1 | 2 - 2 | 232 | 75 | |
XM_017013938.1 | XP_016869427.1 | 2 - 2 | 257 | 75 | |
GenBank mRNA | AK057707.1 | - | 1 - 1 | 152 | 75 |
AK312592.1 | - | 2 - 2 | 170 | 75 | |
BC005850.1 | - | 1 - 1 | 153 | 75 | |
BC067078.1 | - | 1 - 1 | 144 | 75 | |
DA499596.1 | - | 2 - 2 | 181 | 75 | |
DA500281.1 | - | 1 - 1 | 164 | 75 | |
DA503204.1 | - | 2 - 2 | 408 | 75 | |
DA504975.1 | - | 1 - 1 | 228 | 75 | |
DA770917.1 | - | 1 - 1 | 191 | 75 | |
DA774453.1 | - | 1 - 1 | 164 | 75 | |
DA776825.1 | - | 2 - 2 | 257 | 75 | |
DA776968.1 | - | 2 - 2 | 263 | 75 | |
DA795825.1 | - | 2 - 2 | 224 | 75 |