Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135213.1 | NP_001128685.1 | 19 - 19 | 4300 | 58 |
NM_001135214.1 | NP_001128686.1 | 19 - 19 | 4444 | 58 | |
NM_014776.3 | NP_055591.2 | 18 - 18 | 4294 | 58 | |
NM_057169.3 | NP_476510.1 | 20 - 20 | 4534 | 58 | |
NM_057170.3 | NP_476511.1 | 18 - 18 | 4150 | 58 | |
XM_005253997.3 | XP_005254054.1 | 20 - 20 | 4526 | 58 | |
XM_005254000.3 | XP_005254057.1 | 19 - 19 | 4376 | 58 | |
XM_006719707.3 | XP_006719770.1 | 20 - 20 | 4484 | 58 | |
XM_006719708.3 | XP_006719771.1 | 19 - 19 | 4394 | 58 | |
XM_006719709.3 | XP_006719772.1 | 19 - 19 | 4379 | 58 | |
XM_006719712.3 | XP_006719775.1 | 17 - 17 | 4040 | 58 | |
XM_017020258.1 | XP_016875747.1 | 19 - 19 | 4391 | 58 | |
XM_017020259.1 | XP_016875748.1 | 20 - 20 | 4530 | 58 | |
GenBank mRNA | AK057814.1 | - | 1 - 1 | 56 | 58 |
AK090652.1 | - | 1 - 1 | 1780 | 58 | |
AK091325.1 | - | 1 - 1 | 1246 | 58 | |
CR749340.1 | - | 6 - 6 | 3024 | 58 | |
D63482.2 | - | 19 - 19 | 4206 | 58 |