Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49435220-49435638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
UniGene
Hs.485527
Gene Chromosome Location
Chr.6: 49430360-49463328 on Build GRCh38
Transcripts
NM_000255.3 XM_005249143.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4186077 c.2113A>G p.I705V Chr.6: 49435467-49435467 on Build GRCh38
742693 c.2033A>G p.H678R Chr.6: 49435547-49435547 on Build GRCh38
5823418 c.2057T>G p.I686S Chr.6: 49435523-49435523 on Build GRCh38
4854710 c.2112G>A p.V704V Chr.6: 49435468-49435468 on Build GRCh38
6106929 c.2122C>T p.Q708* Chr.6: 49435458-49435458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772442006 Chr.6: 49435539-49435539 on Build GRCh38
rs769217429 Chr.6: 49435457-49435457 on Build GRCh38
rs903031131 Chr.6: 49435567-49435567 on Build GRCh38
rs757366369 Chr.6: 49435606-49435606 on Build GRCh38
rs771886910 Chr.6: 49435451-49435452 on Build GRCh38
rs772652266 Chr.6: 49435558-49435558 on Build GRCh38
rs1007392618 Chr.6: 49435310-49435310 on Build GRCh38
rs751397358 Chr.6: 49435406-49435406 on Build GRCh38
rs757478500 Chr.6: 49435509-49435509 on Build GRCh38
rs888918244 Chr.6: 49435246-49435246 on Build GRCh38
rs149502980 Chr.6: 49435395-49435395 on Build GRCh38
rs147715336 Chr.6: 49435554-49435554 on Build GRCh38
rs565376922 Chr.6: 49435524-49435525 on Build GRCh38
rs985840249 Chr.6: 49435335-49435335 on Build GRCh38
rs8589 Chr.6: 49435569-49435569 on Build GRCh38
rs1017373794 Chr.6: 49435373-49435373 on Build GRCh38
rs767453976 Chr.6: 49435583-49435583 on Build GRCh38
rs756225782 Chr.6: 49435499-49435499 on Build GRCh38
rs121918255 Chr.6: 49435473-49435473 on Build GRCh38
rs543029288 Chr.6: 49435455-49435455 on Build GRCh38
rs777758903 Chr.6: 49435500-49435500 on Build GRCh38
rs916028744 Chr.6: 49435520-49435520 on Build GRCh38
rs776430285 Chr.6: 49435584-49435584 on Build GRCh38
rs578045379 Chr.6: 49435292-49435292 on Build GRCh38
rs879253849 Chr.6: 49435502-49435502 on Build GRCh38
rs373708083 Chr.6: 49435555-49435555 on Build GRCh38
rs201456803 Chr.6: 49435534-49435534 on Build GRCh38
rs200961731 Chr.6: 49435430-49435430 on Build GRCh38
rs751462903 Chr.6: 49435487-49435487 on Build GRCh38
rs879253848 Chr.6: 49435605-49435605 on Build GRCh38
rs761432911 Chr.6: 49435446-49435447 on Build GRCh38
rs974278931 Chr.6: 49435419-49435419 on Build GRCh38
rs376257828 Chr.6: 49435440-49435440 on Build GRCh38
rs760579526 Chr.6: 49435585-49435585 on Build GRCh38
rs147094927 Chr.6: 49435547-49435547 on Build GRCh38
rs367850243 Chr.6: 49435557-49435557 on Build GRCh38
rs1030383808 Chr.6: 49435281-49435281 on Build GRCh38
rs561197473 Chr.6: 49435496-49435496 on Build GRCh38
rs754072258 Chr.6: 49435502-49435502 on Build GRCh38
rs746167463 Chr.6: 49435528-49435528 on Build GRCh38
rs575636400 Chr.6: 49435405-49435405 on Build GRCh38
rs144146728 Chr.6: 49435588-49435588 on Build GRCh38
rs553123132 Chr.6: 49435280-49435280 on Build GRCh38
rs545438554 Chr.6: 49435319-49435319 on Build GRCh38
rs779331475 Chr.6: 49435518-49435518 on Build GRCh38
rs770423282 Chr.6: 49435565-49435565 on Build GRCh38
rs866763981 Chr.6: 49435533-49435533 on Build GRCh38
rs140600746 Chr.6: 49435481-49435481 on Build GRCh38
rs864309739 Chr.6: 49435526-49435526 on Build GRCh38
rs534914349 Chr.6: 49435277-49435277 on Build GRCh38
rs748220054 Chr.6: 49435245-49435245 on Build GRCh38
rs901914392 Chr.6: 49435320-49435320 on Build GRCh38
rs771299021 Chr.6: 49435420-49435420 on Build GRCh38
rs370718165 Chr.6: 49435611-49435611 on Build GRCh38
rs746361551 Chr.6: 49435439-49435439 on Build GRCh38
rs759407117 Chr.6: 49435493-49435493 on Build GRCh38
rs772888575 Chr.6: 49435466-49435466 on Build GRCh38
rs137958217 Chr.6: 49435589-49435589 on Build GRCh38
rs752634070 Chr.6: 49435575-49435575 on Build GRCh38
rs752705830 Chr.6: 49435498-49435498 on Build GRCh38
rs370357524 Chr.6: 49435452-49435452 on Build GRCh38
rs759354207 Chr.6: 49435571-49435571 on Build GRCh38

View Full Product Details