Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16986720-16987216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4025164 c.3074C>T p.A1025V Chr.1: 16986834-16986834 on Build GRCh38
4781110 c.2871G>A p.V957V Chr.1: 16987126-16987126 on Build GRCh38
899712 c.3221C>T p.P1074L Chr.1: 16986819-16986819 on Build GRCh38
4025168 c.2807G>A p.R936H Chr.1: 16987190-16987190 on Build GRCh38
6009108 c.3102G>T p.R1034S Chr.1: 16986938-16986938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779674096 Chr.1: 16986757-16986757 on Build GRCh38
rs767135194 Chr.1: 16986979-16986979 on Build GRCh38
rs758287038 Chr.1: 16986749-16986750 on Build GRCh38
rs137853967 Chr.1: 16986864-16986864 on Build GRCh38
rs750149700 Chr.1: 16987150-16987150 on Build GRCh38
rs779969316 Chr.1: 16987090-16987090 on Build GRCh38
rs745888153 Chr.1: 16986936-16986936 on Build GRCh38
rs9435659 Chr.1: 16986848-16986848 on Build GRCh38
rs531265631 Chr.1: 16987032-16987032 on Build GRCh38
rs763544862 Chr.1: 16987040-16987040 on Build GRCh38
rs751473461 Chr.1: 16987152-16987152 on Build GRCh38
rs371220869 Chr.1: 16986825-16986825 on Build GRCh38
rs567482589 Chr.1: 16987163-16987163 on Build GRCh38
rs759878626 Chr.1: 16987025-16987028 on Build GRCh38
rs944188699 Chr.1: 16986883-16986883 on Build GRCh38
rs747907490 Chr.1: 16986822-16986822 on Build GRCh38
rs199661502 Chr.1: 16986753-16986760 on Build GRCh38
rs377380258 Chr.1: 16986753-16986764 on Build GRCh38
rs538236343 Chr.1: 16986929-16986929 on Build GRCh38
rs56165294 Chr.1: 16987013-16987013 on Build GRCh38
rs367760588 Chr.1: 16987068-16987068 on Build GRCh38
rs762212020 Chr.1: 16986869-16986869 on Build GRCh38
rs768797555 Chr.1: 16987019-16987019 on Build GRCh38
rs764808827 Chr.1: 16987148-16987148 on Build GRCh38
rs150748722 Chr.1: 16987190-16987190 on Build GRCh38
rs56146840 Chr.1: 16986788-16986788 on Build GRCh38
rs140394047 Chr.1: 16986881-16986881 on Build GRCh38
rs146559160 Chr.1: 16987151-16987151 on Build GRCh38
rs747103119 Chr.1: 16987016-16987016 on Build GRCh38
rs773787370 Chr.1: 16986976-16986976 on Build GRCh38
rs369142791 Chr.1: 16986749-16986749 on Build GRCh38
rs772117418 Chr.1: 16986944-16986944 on Build GRCh38
rs367885671 Chr.1: 16987012-16987012 on Build GRCh38
rs376802141 Chr.1: 16987123-16987123 on Build GRCh38
rs755068593 Chr.1: 16987114-16987114 on Build GRCh38
rs571952676 Chr.1: 16987161-16987161 on Build GRCh38
rs55885047 Chr.1: 16987022-16987022 on Build GRCh38
rs373946665 Chr.1: 16986993-16986993 on Build GRCh38
rs767626197 Chr.1: 16987158-16987158 on Build GRCh38
rs763038059 Chr.1: 16986781-16986781 on Build GRCh38
rs765075611 Chr.1: 16987074-16987074 on Build GRCh38
rs200045802 Chr.1: 16986991-16986991 on Build GRCh38
rs56126202 Chr.1: 16986896-16986896 on Build GRCh38
rs746638178 Chr.1: 16986771-16986771 on Build GRCh38
rs773521014 Chr.1: 16987036-16987036 on Build GRCh38
rs761512701 Chr.1: 16987146-16987146 on Build GRCh38
rs757076945 Chr.1: 16986928-16986928 on Build GRCh38
rs958645946 Chr.1: 16987020-16987020 on Build GRCh38
rs763606239 Chr.1: 16987055-16987055 on Build GRCh38
rs764988645 Chr.1: 16986834-16986834 on Build GRCh38
rs751596493 Chr.1: 16987099-16987099 on Build GRCh38
rs777700606 Chr.1: 16986914-16986914 on Build GRCh38
rs143483351 Chr.1: 16986750-16986761 on Build GRCh38
rs764410859 Chr.1: 16986865-16986865 on Build GRCh38
rs777045615 Chr.1: 16986866-16986866 on Build GRCh38
rs892885782 Chr.1: 16986932-16986932 on Build GRCh38
rs145548316 Chr.1: 16987157-16987157 on Build GRCh38
rs771581490 Chr.1: 16987044-16987044 on Build GRCh38
rs951584451 Chr.1: 16986851-16986851 on Build GRCh38
rs202166353 Chr.1: 16987089-16987089 on Build GRCh38
rs374690625 Chr.1: 16987048-16987048 on Build GRCh38
rs752317109 Chr.1: 16986981-16986981 on Build GRCh38
rs372182128 Chr.1: 16987070-16987070 on Build GRCh38
rs773291692 Chr.1: 16986776-16986776 on Build GRCh38
rs766447629 Chr.1: 16986905-16986905 on Build GRCh38
rs976061203 Chr.1: 16986793-16986793 on Build GRCh38
rs777890363 Chr.1: 16987170-16987170 on Build GRCh38
rs138212767 Chr.1: 16987177-16987177 on Build GRCh38
rs148201608 Chr.1: 16987187-16987187 on Build GRCh38
rs774238872 Chr.1: 16986835-16986835 on Build GRCh38
rs375552345 Chr.1: 16987178-16987178 on Build GRCh38
rs760170294 Chr.1: 16987142-16987142 on Build GRCh38
rs971333396 Chr.1: 16986823-16986823 on Build GRCh38
rs137877665 Chr.1: 16986833-16986833 on Build GRCh38
rs749123699 Chr.1: 16986922-16986922 on Build GRCh38
rs768922216 Chr.1: 16986963-16986963 on Build GRCh38
rs769818426 Chr.1: 16986774-16986774 on Build GRCh38
rs184878897 Chr.1: 16986953-16986953 on Build GRCh38
rs369278996 Chr.1: 16986792-16986792 on Build GRCh38
rs974089234 Chr.1: 16986742-16986742 on Build GRCh38
rs779460783 Chr.1: 16987182-16987182 on Build GRCh38
rs373315947 Chr.1: 16986789-16986789 on Build GRCh38
rs116456892 Chr.1: 16986784-16986784 on Build GRCh38
rs377431904 Chr.1: 16986847-16986847 on Build GRCh38
rs376475762 Chr.1: 16986785-16986785 on Build GRCh38
rs372993383 Chr.1: 16986818-16986818 on Build GRCh38
rs1046254851 Chr.1: 16986876-16986876 on Build GRCh38
rs761421 Chr.1: 16987159-16987159 on Build GRCh38
rs867009100 Chr.1: 16987088-16987088 on Build GRCh38
rs777755640 Chr.1: 16986826-16986826 on Build GRCh38
rs1047701202 Chr.1: 16986764-16986764 on Build GRCh38
rs748408338 Chr.1: 16987031-16987031 on Build GRCh38
rs776071534 Chr.1: 16987145-16987145 on Build GRCh38
rs755900033 Chr.1: 16986911-16986911 on Build GRCh38
rs567315420 Chr.1: 16986795-16986795 on Build GRCh38
rs757851198 Chr.1: 16986794-16986794 on Build GRCh38
rs760935912 Chr.1: 16986863-16986863 on Build GRCh38
rs56299881 Chr.1: 16986972-16986972 on Build GRCh38
rs771603791 Chr.1: 16986973-16986973 on Build GRCh38
rs750963832 Chr.1: 16986895-16986895 on Build GRCh38
rs757019364 Chr.1: 16987164-16987164 on Build GRCh38
rs139462252 Chr.1: 16987047-16987047 on Build GRCh38
rs7531163 Chr.1: 16986959-16986959 on Build GRCh38
rs765632065 Chr.1: 16987072-16987072 on Build GRCh38
rs777252902 Chr.1: 16987017-16987018 on Build GRCh38
rs373511503 Chr.1: 16987144-16987144 on Build GRCh38
rs768397313 Chr.1: 16986772-16986772 on Build GRCh38
rs951420437 Chr.1: 16987061-16987061 on Build GRCh38
rs368908107 Chr.1: 16986966-16986966 on Build GRCh38
rs930131952 Chr.1: 16986762-16986762 on Build GRCh38
rs369718815 Chr.1: 16986773-16986773 on Build GRCh38
rs749677780 Chr.1: 16987128-16987128 on Build GRCh38
rs200537435 Chr.1: 16987086-16987086 on Build GRCh38
rs868043973 Chr.1: 16986812-16986812 on Build GRCh38
rs371155708 Chr.1: 16986857-16986857 on Build GRCh38
rs747617559 Chr.1: 16986886-16986887 on Build GRCh38
rs906992159 Chr.1: 16986960-16986960 on Build GRCh38
rs188473562 Chr.1: 16986986-16986986 on Build GRCh38
rs746372968 Chr.1: 16987186-16987186 on Build GRCh38
rs761031832 Chr.1: 16986790-16986790 on Build GRCh38
rs778144698 Chr.1: 16987126-16987126 on Build GRCh38
rs781173666 Chr.1: 16987116-16987116 on Build GRCh38
rs748402756 Chr.1: 16987117-16987117 on Build GRCh38
rs764162255 Chr.1: 16987134-16987134 on Build GRCh38
rs376397035 Chr.1: 16986791-16986791 on Build GRCh38
rs765618139 Chr.1: 16986967-16986967 on Build GRCh38
rs371344553 Chr.1: 16986987-16986987 on Build GRCh38
rs780992643 Chr.1: 16986819-16986819 on Build GRCh38
rs369590596 Chr.1: 16987135-16987135 on Build GRCh38
rs757098756 Chr.1: 16986990-16986990 on Build GRCh38
rs146884019 Chr.1: 16986909-16986909 on Build GRCh38
rs368292341 Chr.1: 16986977-16986977 on Build GRCh38
rs1057519290 Chr.1: 16986886-16986888 on Build GRCh38

View Full Product Details