Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20633603-20634132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6084
Entrez Gene ID
102464833
Gene Name
microRNA 6084
Gene Aliases
hsa-mir-6084
UniGene
Gene Chromosome Location
Chr.1: 20633679-20633788 on Build GRCh38
Transcripts
NR_106732.1 NM_032409.2

Gene Symbol
PINK1
Entrez Gene ID
65018
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
UniGene
Hs.389171
Gene Chromosome Location
Chr.1: 20633455-20651511 on Build GRCh38
Transcripts
NR_106732.1 NM_032409.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143294 c.385C>A p.Q129K Chr.1: 20633933-20633933 on Build GRCh38
3746761 c.189C>T p.L63L Chr.1: 20633737-20633737 on Build GRCh38
5969127 c.161C>A p.A54E Chr.1: 20633709-20633709 on Build GRCh38
7292098 c.329T>G p.L110R Chr.1: 20633877-20633877 on Build GRCh38
5625875 c.314G>T p.G105V Chr.1: 20633862-20633862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941372770 Chr.1: 20633779-20633781 on Build GRCh38
rs765916107 Chr.1: 20633946-20633946 on Build GRCh38
rs537679886 Chr.1: 20633713-20633713 on Build GRCh38
rs771132373 Chr.1: 20633896-20633896 on Build GRCh38
rs368222506 Chr.1: 20633955-20633955 on Build GRCh38
rs1034551549 Chr.1: 20634046-20634046 on Build GRCh38
rs1024029502 Chr.1: 20634029-20634029 on Build GRCh38
rs759968468 Chr.1: 20633732-20633732 on Build GRCh38
rs562271812 Chr.1: 20634008-20634025 on Build GRCh38
rs767084470 Chr.1: 20633950-20633950 on Build GRCh38
rs569753606 Chr.1: 20633636-20633636 on Build GRCh38
rs762327080 Chr.1: 20633941-20633941 on Build GRCh38
rs890987834 Chr.1: 20633855-20633855 on Build GRCh38
rs774802703 Chr.1: 20633731-20633731 on Build GRCh38
rs956491558 Chr.1: 20633721-20633721 on Build GRCh38
rs745900732 Chr.1: 20633911-20633911 on Build GRCh38
rs202048763 Chr.1: 20633766-20633766 on Build GRCh38
rs769273312 Chr.1: 20633968-20633968 on Build GRCh38
rs1051192881 Chr.1: 20634045-20634045 on Build GRCh38
rs918611736 Chr.1: 20634011-20634011 on Build GRCh38
rs574137009 Chr.1: 20633724-20633724 on Build GRCh38
rs777146957 Chr.1: 20633933-20633933 on Build GRCh38
rs752704476 Chr.1: 20633882-20633882 on Build GRCh38
rs776101618 Chr.1: 20633835-20633835 on Build GRCh38
rs555704823 Chr.1: 20633719-20633719 on Build GRCh38
rs775809722 Chr.1: 20633925-20633925 on Build GRCh38
rs866159295 Chr.1: 20633902-20633902 on Build GRCh38
rs766471374 Chr.1: 20633725-20633725 on Build GRCh38
rs762698585 Chr.1: 20633859-20633859 on Build GRCh38
rs869033002 Chr.1: 20633903-20633903 on Build GRCh38
rs1001702223 Chr.1: 20634042-20634042 on Build GRCh38
rs1018195590 Chr.1: 20633886-20633886 on Build GRCh38
rs767869808 Chr.1: 20633734-20633734 on Build GRCh38
rs971528785 Chr.1: 20633860-20633860 on Build GRCh38
rs772448788 Chr.1: 20633828-20633828 on Build GRCh38
rs866153120 Chr.1: 20633772-20633772 on Build GRCh38
rs979028448 Chr.1: 20633707-20633707 on Build GRCh38
rs770933972 Chr.1: 20633815-20633815 on Build GRCh38
rs779265931 Chr.1: 20633907-20633907 on Build GRCh38
rs974316684 Chr.1: 20634006-20634006 on Build GRCh38
rs569642671 Chr.1: 20634065-20634065 on Build GRCh38
rs1007114265 Chr.1: 20633885-20633885 on Build GRCh38
rs751445683 Chr.1: 20633773-20633773 on Build GRCh38
rs1038345456 Chr.1: 20633784-20633784 on Build GRCh38
rs751456355 Chr.1: 20633717-20633718 on Build GRCh38
rs1057519104 Chr.1: 20633717-20633717 on Build GRCh38
rs781710561 Chr.1: 20634083-20634083 on Build GRCh38
rs774312092 Chr.1: 20633817-20633817 on Build GRCh38
rs867771554 Chr.1: 20633709-20633709 on Build GRCh38
rs117438827 Chr.1: 20633949-20633949 on Build GRCh38
rs918623975 Chr.1: 20633917-20633917 on Build GRCh38
rs754557745 Chr.1: 20633878-20633878 on Build GRCh38
rs547423601 Chr.1: 20633954-20633954 on Build GRCh38
rs780970528 Chr.1: 20633881-20633881 on Build GRCh38
rs967760539 Chr.1: 20633701-20633701 on Build GRCh38
rs944507113 Chr.1: 20633942-20633942 on Build GRCh38
rs761293056 Chr.1: 20633932-20633932 on Build GRCh38
rs781257771 Chr.1: 20633776-20633776 on Build GRCh38
rs769626575 Chr.1: 20633793-20633793 on Build GRCh38
rs763142730 Chr.1: 20633747-20633747 on Build GRCh38
rs563467429 Chr.1: 20633810-20633810 on Build GRCh38
rs963276391 Chr.1: 20634064-20634064 on Build GRCh38
rs956355350 Chr.1: 20633947-20633947 on Build GRCh38
rs897150702 Chr.1: 20633786-20633786 on Build GRCh38
rs879182315 Chr.1: 20633970-20633970 on Build GRCh38
rs372834836 Chr.1: 20634017-20634034 on Build GRCh38
rs779984827 Chr.1: 20633717-20633749 on Build GRCh38
rs942558477 Chr.1: 20634047-20634048 on Build GRCh38
rs1010332507 Chr.1: 20633983-20633983 on Build GRCh38
rs748343847 Chr.1: 20633708-20633708 on Build GRCh38
rs777344911 Chr.1: 20633975-20633975 on Build GRCh38
rs747462269 Chr.1: 20633966-20633966 on Build GRCh38
rs751529167 Chr.1: 20633914-20633914 on Build GRCh38
rs148871409 Chr.1: 20633892-20633892 on Build GRCh38
rs575668171 Chr.1: 20633840-20633840 on Build GRCh38
rs951384154 Chr.1: 20634098-20634098 on Build GRCh38
rs752507877 Chr.1: 20633953-20633953 on Build GRCh38
rs759723977 Chr.1: 20633819-20633819 on Build GRCh38
rs749546463 Chr.1: 20633814-20633814 on Build GRCh38
rs758372183 Chr.1: 20633962-20633962 on Build GRCh38
rs377219724 Chr.1: 20633960-20633960 on Build GRCh38
rs753621873 Chr.1: 20633959-20633959 on Build GRCh38
rs747865765 Chr.1: 20633787-20633787 on Build GRCh38
rs751020216 Chr.1: 20633874-20633874 on Build GRCh38
rs1051163566 Chr.1: 20633926-20633926 on Build GRCh38
rs551375519 Chr.1: 20634032-20634032 on Build GRCh38
rs751273819 Chr.1: 20633948-20633948 on Build GRCh38
rs758019868 Chr.1: 20633662-20633662 on Build GRCh38
rs780188375 Chr.1: 20633964-20633964 on Build GRCh38
rs755000580 Chr.1: 20633821-20633821 on Build GRCh38
rs1032715657 Chr.1: 20633804-20633804 on Build GRCh38
rs746940423 Chr.1: 20633687-20633687 on Build GRCh38
rs763000118 Chr.1: 20633723-20633723 on Build GRCh38
rs1006830855 Chr.1: 20633649-20633649 on Build GRCh38
rs757930144 Chr.1: 20633764-20633764 on Build GRCh38
rs1010013404 Chr.1: 20633833-20633833 on Build GRCh38
rs544984102 Chr.1: 20633796-20633796 on Build GRCh38
rs764573020 Chr.1: 20633848-20633848 on Build GRCh38
rs577010119 Chr.1: 20633641-20633641 on Build GRCh38
rs987212439 Chr.1: 20634100-20634100 on Build GRCh38
rs750054121 Chr.1: 20633851-20633851 on Build GRCh38
rs772239179 Chr.1: 20633916-20633916 on Build GRCh38
rs768456559 Chr.1: 20633693-20633693 on Build GRCh38
rs780115582 Chr.1: 20633668-20633668 on Build GRCh38
rs867839970 Chr.1: 20633934-20633934 on Build GRCh38
rs45530340 Chr.1: 20633737-20633737 on Build GRCh38
rs776293086 Chr.1: 20633703-20633703 on Build GRCh38
rs553247784 Chr.1: 20633744-20633744 on Build GRCh38
rs750146116 Chr.1: 20633761-20633761 on Build GRCh38
rs543071128 Chr.1: 20633862-20633862 on Build GRCh38
rs866876745 Chr.1: 20633760-20633760 on Build GRCh38
rs754884141 Chr.1: 20633774-20633774 on Build GRCh38
rs749456905 Chr.1: 20633893-20633893 on Build GRCh38

View Full Product Details