Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20651113-20651620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PINK1
Entrez Gene ID
65018
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
UniGene
Hs.389171
Gene Chromosome Location
Chr.1: 20633455-20651511 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Gene Symbol
PINK1-AS
Entrez Gene ID
100861548
Gene Name
PINK1 antisense RNA
Gene Aliases
NAPINK1, PINK1-AS1, PINK1AS
UniGene
Hs.710259
Gene Chromosome Location
Chr.1: 20642657-20652193 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

SNP Detail

SNP ID Chromosome Location
rs890606959 Chr.1: 20651497-20651497 on Build GRCh38
rs994768322 Chr.1: 20651479-20651479 on Build GRCh38
rs917175167 Chr.1: 20651147-20651147 on Build GRCh38
rs1000295586 Chr.1: 20651373-20651373 on Build GRCh38
rs543456950 Chr.1: 20651295-20651295 on Build GRCh38
rs1027341650 Chr.1: 20651304-20651304 on Build GRCh38
rs975014860 Chr.1: 20651378-20651378 on Build GRCh38
rs555622542 Chr.1: 20651575-20651575 on Build GRCh38
rs1020758910 Chr.1: 20651229-20651229 on Build GRCh38
rs1038492908 Chr.1: 20651224-20651225 on Build GRCh38
rs190393061 Chr.1: 20651223-20651223 on Build GRCh38
rs886045851 Chr.1: 20651262-20651262 on Build GRCh38
rs181532922 Chr.1: 20651408-20651408 on Build GRCh38
rs990371699 Chr.1: 20651571-20651571 on Build GRCh38
rs146899464 Chr.1: 20651595-20651595 on Build GRCh38
rs950417953 Chr.1: 20651323-20651323 on Build GRCh38
rs1054458936 Chr.1: 20651210-20651210 on Build GRCh38
rs533348669 Chr.1: 20651425-20651425 on Build GRCh38
rs375956041 Chr.1: 20651267-20651268 on Build GRCh38
rs979749634 Chr.1: 20651193-20651193 on Build GRCh38
rs184877436 Chr.1: 20651277-20651277 on Build GRCh38
rs913230569 Chr.1: 20651579-20651579 on Build GRCh38
rs757779349 Chr.1: 20651523-20651523 on Build GRCh38
rs537315041 Chr.1: 20651482-20651482 on Build GRCh38
rs758748349 Chr.1: 20651512-20651512 on Build GRCh38
rs189715142 Chr.1: 20651535-20651535 on Build GRCh38
rs983219356 Chr.1: 20651518-20651518 on Build GRCh38
rs752692312 Chr.1: 20651432-20651432 on Build GRCh38
rs572231570 Chr.1: 20651549-20651549 on Build GRCh38
rs78349864 Chr.1: 20651498-20651498 on Build GRCh38
rs1019144290 Chr.1: 20651529-20651529 on Build GRCh38
rs141140099 Chr.1: 20651346-20651346 on Build GRCh38
rs1003457823 Chr.1: 20651262-20651262 on Build GRCh38
rs34119921 Chr.1: 20651413-20651414 on Build GRCh38
rs555315482 Chr.1: 20651186-20651186 on Build GRCh38
rs556390159 Chr.1: 20651261-20651261 on Build GRCh38
rs398069438 Chr.1: 20651419-20651420 on Build GRCh38
rs971169305 Chr.1: 20651585-20651585 on Build GRCh38
rs912025937 Chr.1: 20651355-20651355 on Build GRCh38
rs34361349 Chr.1: 20651414-20651415 on Build GRCh38
rs113674347 Chr.1: 20651427-20651428 on Build GRCh38
rs1054363642 Chr.1: 20651399-20651399 on Build GRCh38
rs886045850 Chr.1: 20651201-20651201 on Build GRCh38
rs570038599 Chr.1: 20651480-20651480 on Build GRCh38
rs896895231 Chr.1: 20651245-20651246 on Build GRCh38
rs941056833 Chr.1: 20651441-20651442 on Build GRCh38
rs546151634 Chr.1: 20651238-20651238 on Build GRCh38
rs112126165 Chr.1: 20651318-20651318 on Build GRCh38
rs650616 Chr.1: 20651565-20651565 on Build GRCh38
rs3077908 Chr.1: 20651366-20651367 on Build GRCh38
rs138181538 Chr.1: 20651218-20651221 on Build GRCh38
rs895040725 Chr.1: 20651448-20651448 on Build GRCh38
rs553977557 Chr.1: 20651225-20651225 on Build GRCh38
rs1048375477 Chr.1: 20651486-20651486 on Build GRCh38
rs112009066 Chr.1: 20651563-20651564 on Build GRCh38
rs185791919 Chr.1: 20651428-20651428 on Build GRCh38
rs758797319 Chr.1: 20651428-20651431 on Build GRCh38
rs3077906 Chr.1: 20651232-20651233 on Build GRCh38
rs1003813219 Chr.1: 20651426-20651426 on Build GRCh38
rs891811639 Chr.1: 20651400-20651400 on Build GRCh38
rs973089088 Chr.1: 20651292-20651292 on Build GRCh38
rs946094086 Chr.1: 20651402-20651402 on Build GRCh38
rs11554544 Chr.1: 20651258-20651258 on Build GRCh38
rs1039271817 Chr.1: 20651505-20651505 on Build GRCh38
rs369053717 Chr.1: 20651177-20651177 on Build GRCh38
rs1043502 Chr.1: 20651255-20651255 on Build GRCh38
rs558718255 Chr.1: 20651316-20651316 on Build GRCh38
rs2078073 Chr.1: 20651363-20651363 on Build GRCh38
rs574097463 Chr.1: 20651200-20651200 on Build GRCh38
rs16824318 Chr.1: 20651181-20651181 on Build GRCh38
rs959054690 Chr.1: 20651279-20651279 on Build GRCh38
rs143763511 Chr.1: 20651261-20651262 on Build GRCh38
rs33999970 Chr.1: 20651361-20651362 on Build GRCh38
rs900466604 Chr.1: 20651370-20651370 on Build GRCh38
rs567546379 Chr.1: 20651502-20651502 on Build GRCh38
rs559936998 Chr.1: 20651420-20651420 on Build GRCh38
rs193299196 Chr.1: 20651236-20651236 on Build GRCh38
rs922209796 Chr.1: 20651393-20651393 on Build GRCh38
rs900498309 Chr.1: 20651201-20651201 on Build GRCh38
rs765673394 Chr.1: 20651209-20651209 on Build GRCh38
rs983595389 Chr.1: 20651163-20651163 on Build GRCh38
rs1573132 Chr.1: 20651531-20651531 on Build GRCh38
rs988656615 Chr.1: 20651144-20651144 on Build GRCh38
rs372726829 Chr.1: 20651441-20651442 on Build GRCh38
rs71760582 Chr.1: 20651364-20651365 on Build GRCh38
rs886045852 Chr.1: 20651304-20651304 on Build GRCh38
rs929729176 Chr.1: 20651485-20651485 on Build GRCh38
rs935987817 Chr.1: 20651396-20651396 on Build GRCh38

View Full Product Details