Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67231743-67232239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749697 c.2047A>G p.K683E Chr.16: 67232194-67232194 on Build GRCh38
5457997 c.2225C>T p.T742M Chr.16: 67231797-67231797 on Build GRCh38
6079528 c.2255C>T p.A752V Chr.16: 67231767-67231767 on Build GRCh38
5677710 c.2227G>C p.E743Q Chr.16: 67231795-67231795 on Build GRCh38
5649250 c.2047-1G>T p.? Chr.16: 67232195-67232195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371242052 Chr.16: 67232218-67232218 on Build GRCh38
rs750309031 Chr.16: 67232174-67232174 on Build GRCh38
rs779699758 Chr.16: 67232100-67232100 on Build GRCh38
rs561172471 Chr.16: 67231980-67231980 on Build GRCh38
rs375464202 Chr.16: 67231990-67231990 on Build GRCh38
rs750497587 Chr.16: 67231797-67231797 on Build GRCh38
rs769500535 Chr.16: 67231872-67231872 on Build GRCh38
rs762830872 Chr.16: 67232022-67232022 on Build GRCh38
rs774413028 Chr.16: 67231991-67231993 on Build GRCh38
rs144361447 Chr.16: 67232138-67232138 on Build GRCh38
rs138444737 Chr.16: 67232124-67232124 on Build GRCh38
rs764489886 Chr.16: 67232212-67232212 on Build GRCh38
rs370261438 Chr.16: 67232175-67232175 on Build GRCh38
rs375932243 Chr.16: 67232064-67232064 on Build GRCh38
rs766696745 Chr.16: 67231803-67231803 on Build GRCh38
rs754625636 Chr.16: 67232104-67232104 on Build GRCh38
rs868302613 Chr.16: 67232043-67232043 on Build GRCh38
rs748467203 Chr.16: 67232217-67232217 on Build GRCh38
rs773788569 Chr.16: 67232137-67232137 on Build GRCh38
rs751343080 Chr.16: 67232044-67232044 on Build GRCh38
rs79286145 Chr.16: 67232018-67232018 on Build GRCh38
rs148737699 Chr.16: 67232155-67232155 on Build GRCh38
rs779613215 Chr.16: 67231927-67231927 on Build GRCh38
rs763448301 Chr.16: 67231930-67231930 on Build GRCh38
rs778622827 Chr.16: 67232216-67232216 on Build GRCh38
rs111290083 Chr.16: 67231856-67231856 on Build GRCh38
rs919044428 Chr.16: 67232131-67232131 on Build GRCh38
rs768849841 Chr.16: 67232003-67232003 on Build GRCh38
rs149612162 Chr.16: 67232126-67232126 on Build GRCh38
rs769206751 Chr.16: 67231772-67231772 on Build GRCh38
rs751407193 Chr.16: 67231843-67231843 on Build GRCh38
rs370312254 Chr.16: 67231978-67231978 on Build GRCh38
rs981586641 Chr.16: 67232073-67232073 on Build GRCh38
rs34888644 Chr.16: 67232162-67232162 on Build GRCh38
rs748898176 Chr.16: 67232102-67232102 on Build GRCh38
rs778120536 Chr.16: 67231860-67231866 on Build GRCh38
rs761974488 Chr.16: 67231786-67231786 on Build GRCh38
rs200917054 Chr.16: 67231814-67231814 on Build GRCh38
rs763761384 Chr.16: 67232029-67232029 on Build GRCh38
rs755558730 Chr.16: 67232096-67232096 on Build GRCh38
rs777779985 Chr.16: 67231837-67231837 on Build GRCh38
rs761598605 Chr.16: 67232163-67232163 on Build GRCh38
rs756826518 Chr.16: 67231844-67231844 on Build GRCh38
rs754056591 Chr.16: 67232021-67232021 on Build GRCh38
rs764644193 Chr.16: 67231972-67231972 on Build GRCh38
rs933180964 Chr.16: 67231918-67231918 on Build GRCh38
rs773482015 Chr.16: 67232016-67232016 on Build GRCh38
rs918990283 Chr.16: 67231886-67231886 on Build GRCh38
rs760044647 Chr.16: 67232007-67232008 on Build GRCh38
rs772521918 Chr.16: 67232130-67232130 on Build GRCh38
rs753364765 Chr.16: 67232179-67232179 on Build GRCh38
rs752607604 Chr.16: 67231805-67231805 on Build GRCh38
rs374165476 Chr.16: 67232176-67232176 on Build GRCh38
rs1015903392 Chr.16: 67232045-67232045 on Build GRCh38
rs767854176 Chr.16: 67232030-67232031 on Build GRCh38
rs945393606 Chr.16: 67232205-67232205 on Build GRCh38
rs760909642 Chr.16: 67232099-67232099 on Build GRCh38
rs745363122 Chr.16: 67231855-67231855 on Build GRCh38
rs767455160 Chr.16: 67231796-67231796 on Build GRCh38
rs780610608 Chr.16: 67231848-67231848 on Build GRCh38
rs775946395 Chr.16: 67232075-67232075 on Build GRCh38
rs772608403 Chr.16: 67232011-67232011 on Build GRCh38
rs765913254 Chr.16: 67232178-67232178 on Build GRCh38
rs1033811337 Chr.16: 67232015-67232015 on Build GRCh38
rs932370623 Chr.16: 67232199-67232199 on Build GRCh38
rs774658881 Chr.16: 67231785-67231785 on Build GRCh38
rs777028123 Chr.16: 67232145-67232145 on Build GRCh38
rs767451584 Chr.16: 67232173-67232173 on Build GRCh38
rs372426679 Chr.16: 67232125-67232125 on Build GRCh38
rs761025263 Chr.16: 67231799-67231799 on Build GRCh38
rs774952608 Chr.16: 67232008-67232008 on Build GRCh38
rs759719315 Chr.16: 67232146-67232146 on Build GRCh38
rs749936714 Chr.16: 67232088-67232088 on Build GRCh38
rs758877940 Chr.16: 67231828-67231828 on Build GRCh38
rs754644846 Chr.16: 67232204-67232204 on Build GRCh38
rs573024809 Chr.16: 67231984-67231984 on Build GRCh38
rs757193319 Chr.16: 67232046-67232046 on Build GRCh38
rs368135843 Chr.16: 67232009-67232009 on Build GRCh38

View Full Product Details