Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67233997-67234491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5013028 c.1637T>C p.F546S Chr.16: 67234066-67234066 on Build GRCh38
3818252 c.1587C>G p.I529M Chr.16: 67234116-67234116 on Build GRCh38
4684673 c.1682_1683delAG p.E561fs*55 Chr.16: 67234020-67234021 on Build GRCh38
4684674 c.1604G>A p.R535H Chr.16: 67234099-67234099 on Build GRCh38
5740689 c.1670T>C p.M557T Chr.16: 67234033-67234033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2271290 Chr.16: 67234123-67234123 on Build GRCh38
rs754761714 Chr.16: 67234261-67234261 on Build GRCh38
rs749275536 Chr.16: 67234038-67234038 on Build GRCh38
rs968974323 Chr.16: 67234138-67234138 on Build GRCh38
rs775216754 Chr.16: 67234432-67234432 on Build GRCh38
rs779755764 Chr.16: 67234467-67234467 on Build GRCh38
rs747604950 Chr.16: 67234076-67234076 on Build GRCh38
rs917660510 Chr.16: 67234180-67234180 on Build GRCh38
rs774862376 Chr.16: 67234412-67234412 on Build GRCh38
rs2271291 Chr.16: 67234246-67234246 on Build GRCh38
rs752536609 Chr.16: 67234279-67234279 on Build GRCh38
rs535833328 Chr.16: 67234376-67234376 on Build GRCh38
rs763415139 Chr.16: 67234349-67234349 on Build GRCh38
rs748075433 Chr.16: 67234418-67234418 on Build GRCh38
rs761027309 Chr.16: 67234422-67234422 on Build GRCh38
rs369969027 Chr.16: 67234178-67234178 on Build GRCh38
rs772681218 Chr.16: 67234089-67234089 on Build GRCh38
rs755729329 Chr.16: 67234464-67234464 on Build GRCh38
rs764393738 Chr.16: 67234129-67234129 on Build GRCh38
rs769190232 Chr.16: 67234350-67234350 on Build GRCh38
rs746516526 Chr.16: 67234291-67234291 on Build GRCh38
rs931128160 Chr.16: 67234193-67234193 on Build GRCh38
rs551923090 Chr.16: 67234219-67234219 on Build GRCh38
rs150240690 Chr.16: 67234390-67234390 on Build GRCh38
rs550457905 Chr.16: 67234099-67234099 on Build GRCh38
rs375228741 Chr.16: 67234308-67234308 on Build GRCh38
rs574692248 Chr.16: 67234361-67234361 on Build GRCh38
rs762878368 Chr.16: 67234112-67234112 on Build GRCh38
rs753670739 Chr.16: 67234144-67234144 on Build GRCh38
rs745904027 Chr.16: 67234300-67234300 on Build GRCh38
rs771435032 Chr.16: 67234177-67234177 on Build GRCh38
rs774427401 Chr.16: 67234199-67234200 on Build GRCh38
rs748259381 Chr.16: 67234416-67234416 on Build GRCh38
rs774359056 Chr.16: 67234353-67234353 on Build GRCh38
rs201535041 Chr.16: 67234156-67234156 on Build GRCh38
rs142023201 Chr.16: 67234377-67234377 on Build GRCh38
rs1008214080 Chr.16: 67234082-67234082 on Build GRCh38
rs371069606 Chr.16: 67234454-67234454 on Build GRCh38
rs775806091 Chr.16: 67234200-67234200 on Build GRCh38
rs148095513 Chr.16: 67234407-67234407 on Build GRCh38
rs878912039 Chr.16: 67234230-67234230 on Build GRCh38
rs1036496181 Chr.16: 67234031-67234031 on Build GRCh38
rs752514725 Chr.16: 67234371-67234371 on Build GRCh38
rs774059853 Chr.16: 67234061-67234061 on Build GRCh38
rs755736544 Chr.16: 67234400-67234400 on Build GRCh38
rs138899852 Chr.16: 67234391-67234391 on Build GRCh38
rs753689172 Chr.16: 67234250-67234250 on Build GRCh38
rs759435821 Chr.16: 67234367-67234367 on Build GRCh38
rs776099857 Chr.16: 67234333-67234333 on Build GRCh38
rs1049360138 Chr.16: 67234381-67234381 on Build GRCh38
rs751687594 Chr.16: 67234136-67234136 on Build GRCh38
rs772669711 Chr.16: 67234399-67234400 on Build GRCh38
rs766612246 Chr.16: 67234429-67234429 on Build GRCh38
rs199614307 Chr.16: 67234262-67234262 on Build GRCh38
rs947794746 Chr.16: 67234184-67234184 on Build GRCh38
rs996907960 Chr.16: 67234280-67234280 on Build GRCh38
rs149368972 Chr.16: 67234421-67234421 on Build GRCh38
rs553913184 Chr.16: 67234387-67234387 on Build GRCh38
rs146941627 Chr.16: 67234370-67234370 on Build GRCh38
rs373647547 Chr.16: 67234165-67234165 on Build GRCh38
rs966095989 Chr.16: 67234358-67234358 on Build GRCh38
rs1055473456 Chr.16: 67234147-67234147 on Build GRCh38
rs758325596 Chr.16: 67234282-67234282 on Build GRCh38
rs770549814 Chr.16: 67234183-67234183 on Build GRCh38
rs74771707 Chr.16: 67234186-67234186 on Build GRCh38
rs59149041 Chr.16: 67234306-67234306 on Build GRCh38
rs756795171 Chr.16: 67234450-67234450 on Build GRCh38
rs1021823768 Chr.16: 67234297-67234297 on Build GRCh38
rs780848946 Chr.16: 67234394-67234394 on Build GRCh38
rs146332890 Chr.16: 67234448-67234448 on Build GRCh38
rs953442121 Chr.16: 67234301-67234301 on Build GRCh38
rs977646364 Chr.16: 67234141-67234141 on Build GRCh38
rs749881387 Chr.16: 67234457-67234457 on Build GRCh38
rs200444812 Chr.16: 67234083-67234083 on Build GRCh38
rs1025618348 Chr.16: 67234032-67234032 on Build GRCh38
rs911986846 Chr.16: 67234337-67234337 on Build GRCh38
rs1020091571 Chr.16: 67234188-67234188 on Build GRCh38
rs1032374984 Chr.16: 67234310-67234310 on Build GRCh38
rs199754555 Chr.16: 67234435-67234435 on Build GRCh38
rs930934962 Chr.16: 67234086-67234086 on Build GRCh38
rs77281081 Chr.16: 67234204-67234204 on Build GRCh38
rs374644316 Chr.16: 67234110-67234110 on Build GRCh38
rs770639558 Chr.16: 67234303-67234303 on Build GRCh38
rs777197560 Chr.16: 67234288-67234288 on Build GRCh38
rs904601743 Chr.16: 67234468-67234468 on Build GRCh38
rs751632193 Chr.16: 67234203-67234203 on Build GRCh38
rs751453077 Chr.16: 67234449-67234449 on Build GRCh38
rs765722561 Chr.16: 67234107-67234107 on Build GRCh38
rs77958237 Chr.16: 67234105-67234105 on Build GRCh38
rs780166979 Chr.16: 67234025-67234025 on Build GRCh38
rs576712879 Chr.16: 67234411-67234411 on Build GRCh38
rs755811201 Chr.16: 67234022-67234022 on Build GRCh38
rs771758797 Chr.16: 67234081-67234081 on Build GRCh38
rs564483976 Chr.16: 67234187-67234187 on Build GRCh38
rs924870207 Chr.16: 67234149-67234149 on Build GRCh38
rs749656540 Chr.16: 67234408-67234408 on Build GRCh38
rs992241696 Chr.16: 67234360-67234360 on Build GRCh38
rs778872929 Chr.16: 67234356-67234356 on Build GRCh38
rs568216912 Chr.16: 67234396-67234396 on Build GRCh38
rs1041457164 Chr.16: 67234440-67234440 on Build GRCh38
rs187388529 Chr.16: 67234319-67234319 on Build GRCh38
rs764116221 Chr.16: 67234201-67234201 on Build GRCh38
rs147764255 Chr.16: 67234154-67234154 on Build GRCh38
rs768690449 Chr.16: 67234040-67234040 on Build GRCh38
rs149055501 Chr.16: 67234092-67234092 on Build GRCh38
rs202036259 Chr.16: 67234142-67234142 on Build GRCh38
rs373548373 Chr.16: 67234365-67234365 on Build GRCh38

View Full Product Details