Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7689142-7689641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
UniGene
Hs.408312
Gene Chromosome Location
Chr.17: 7686071-7703502 on Build GRCh38
Transcripts
NM_001143990.1 NM_001143991.1 NM_001143992.1 NM_018081.2 XM_011523952.2 XM_017024812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6082217 c.495C>T p.S165S Chr.17: 7689287-7689287 on Build GRCh38
984997 c.540C>T p.D180D Chr.17: 7689599-7689599 on Build GRCh38
3421940 c.432C>T p.T144T Chr.17: 7689224-7689224 on Build GRCh38
148182 c.450C>T p.F150F Chr.17: 7689242-7689242 on Build GRCh38
6016467 c.480C>T p.S160S Chr.17: 7689272-7689272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903031697 Chr.17: 7689562-7689562 on Build GRCh38
rs972224971 Chr.17: 7689391-7689391 on Build GRCh38
rs926446428 Chr.17: 7689535-7689535 on Build GRCh38
rs770992500 Chr.17: 7689288-7689288 on Build GRCh38
rs749881818 Chr.17: 7689342-7689342 on Build GRCh38
rs776832907 Chr.17: 7689609-7689609 on Build GRCh38
rs536866484 Chr.17: 7689507-7689507 on Build GRCh38
rs369563538 Chr.17: 7689237-7689237 on Build GRCh38
rs374218289 Chr.17: 7689340-7689340 on Build GRCh38
rs951141640 Chr.17: 7689446-7689446 on Build GRCh38
rs967950820 Chr.17: 7689512-7689512 on Build GRCh38
rs539554212 Chr.17: 7689311-7689311 on Build GRCh38
rs768560508 Chr.17: 7689184-7689184 on Build GRCh38
rs773340183 Chr.17: 7689201-7689201 on Build GRCh38
rs781128964 Chr.17: 7689372-7689372 on Build GRCh38
rs147817526 Chr.17: 7689287-7689287 on Build GRCh38
rs140594263 Chr.17: 7689226-7689226 on Build GRCh38
rs979774047 Chr.17: 7689534-7689534 on Build GRCh38
rs2287498 Chr.17: 7689242-7689242 on Build GRCh38
rs750056345 Chr.17: 7689244-7689244 on Build GRCh38
rs753461311 Chr.17: 7689238-7689238 on Build GRCh38
rs281865547 Chr.17: 7689284-7689284 on Build GRCh38
rs769107489 Chr.17: 7689603-7689603 on Build GRCh38
rs764697968 Chr.17: 7689338-7689338 on Build GRCh38
rs147226406 Chr.17: 7689225-7689225 on Build GRCh38
rs746794861 Chr.17: 7689261-7689261 on Build GRCh38
rs887378979 Chr.17: 7689168-7689168 on Build GRCh38
rs928415437 Chr.17: 7689469-7689469 on Build GRCh38
rs771063284 Chr.17: 7689209-7689209 on Build GRCh38
rs35345788 Chr.17: 7689501-7689502 on Build GRCh38
rs761113077 Chr.17: 7689336-7689336 on Build GRCh38
rs955495095 Chr.17: 7689478-7689478 on Build GRCh38
rs570571157 Chr.17: 7689442-7689442 on Build GRCh38
rs774574319 Chr.17: 7689211-7689211 on Build GRCh38
rs376032429 Chr.17: 7689555-7689555 on Build GRCh38
rs772485427 Chr.17: 7689570-7689570 on Build GRCh38
rs947311558 Chr.17: 7689533-7689533 on Build GRCh38
rs565530006 Chr.17: 7689263-7689263 on Build GRCh38
rs761263963 Chr.17: 7689339-7689339 on Build GRCh38
rs17882137 Chr.17: 7689202-7689202 on Build GRCh38
rs374862663 Chr.17: 7689370-7689370 on Build GRCh38
rs766017619 Chr.17: 7689352-7689352 on Build GRCh38
rs779875361 Chr.17: 7689258-7689258 on Build GRCh38
rs936989880 Chr.17: 7689482-7689482 on Build GRCh38
rs141837480 Chr.17: 7689457-7689457 on Build GRCh38
rs1029683725 Chr.17: 7689476-7689476 on Build GRCh38
rs755972975 Chr.17: 7689552-7689552 on Build GRCh38
rs747404789 Chr.17: 7689602-7689602 on Build GRCh38
rs2287497 Chr.17: 7689462-7689462 on Build GRCh38
rs964128624 Chr.17: 7689207-7689207 on Build GRCh38
rs199587902 Chr.17: 7689329-7689329 on Build GRCh38
rs547410263 Chr.17: 7689371-7689371 on Build GRCh38
rs779095345 Chr.17: 7689554-7689554 on Build GRCh38
rs377154284 Chr.17: 7689347-7689347 on Build GRCh38
rs751360976 Chr.17: 7689353-7689353 on Build GRCh38
rs780223030 Chr.17: 7689360-7689360 on Build GRCh38
rs750964508 Chr.17: 7689196-7689196 on Build GRCh38
rs772847669 Chr.17: 7689617-7689617 on Build GRCh38
rs571153185 Chr.17: 7689249-7689249 on Build GRCh38
rs765650726 Chr.17: 7689610-7689610 on Build GRCh38
rs992019159 Chr.17: 7689556-7689556 on Build GRCh38
rs937718476 Chr.17: 7689310-7689310 on Build GRCh38
rs774466254 Chr.17: 7689294-7689294 on Build GRCh38
rs1005714403 Chr.17: 7689169-7689169 on Build GRCh38
rs758386953 Chr.17: 7689283-7689284 on Build GRCh38
rs776051850 Chr.17: 7689330-7689330 on Build GRCh38
rs17886268 Chr.17: 7689484-7689484 on Build GRCh38
rs748077742 Chr.17: 7689541-7689541 on Build GRCh38
rs764681311 Chr.17: 7689234-7689234 on Build GRCh38
rs759839550 Chr.17: 7689221-7689221 on Build GRCh38
rs781041136 Chr.17: 7689271-7689271 on Build GRCh38
rs746990787 Chr.17: 7689177-7689177 on Build GRCh38
rs747235125 Chr.17: 7689549-7689550 on Build GRCh38
rs775784125 Chr.17: 7689599-7689599 on Build GRCh38
rs1017552747 Chr.17: 7689170-7689170 on Build GRCh38
rs17885803 Chr.17: 7689164-7689164 on Build GRCh38

View Full Product Details