Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35575083-35575609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
UniGene
Hs.591190
Gene Chromosome Location
Chr.17: 35574795-35578637 on Build GRCh38
Transcripts
NM_000286.2

SNP Detail

SNP ID Chromosome Location
rs529066074 Chr.17: 35575244-35575244 on Build GRCh38
rs1004771102 Chr.17: 35575283-35575283 on Build GRCh38
rs113891712 Chr.17: 35575485-35575485 on Build GRCh38
rs147182821 Chr.17: 35575230-35575232 on Build GRCh38
rs3031851 Chr.17: 35575212-35575213 on Build GRCh38
rs886052827 Chr.17: 35575263-35575263 on Build GRCh38
rs892593344 Chr.17: 35575162-35575162 on Build GRCh38
rs989722936 Chr.17: 35575466-35575466 on Build GRCh38
rs1046321 Chr.17: 35575265-35575265 on Build GRCh38
rs547399279 Chr.17: 35575353-35575353 on Build GRCh38
rs1016608839 Chr.17: 35575349-35575349 on Build GRCh38
rs541598688 Chr.17: 35575213-35575213 on Build GRCh38
rs539431314 Chr.17: 35575111-35575111 on Build GRCh38
rs1010846026 Chr.17: 35575211-35575211 on Build GRCh38
rs762716335 Chr.17: 35575402-35575402 on Build GRCh38
rs750063724 Chr.17: 35575266-35575266 on Build GRCh38
rs1034683504 Chr.17: 35575240-35575240 on Build GRCh38
rs1045152970 Chr.17: 35575264-35575264 on Build GRCh38
rs886052826 Chr.17: 35575250-35575250 on Build GRCh38
rs895745225 Chr.17: 35575525-35575525 on Build GRCh38
rs948237244 Chr.17: 35575263-35575263 on Build GRCh38
rs570189667 Chr.17: 35575554-35575554 on Build GRCh38
rs570468178 Chr.17: 35575425-35575425 on Build GRCh38
rs577816577 Chr.17: 35575112-35575112 on Build GRCh38
rs905163839 Chr.17: 35575484-35575484 on Build GRCh38
rs543539786 Chr.17: 35575129-35575129 on Build GRCh38
rs1003538955 Chr.17: 35575270-35575270 on Build GRCh38
rs887110828 Chr.17: 35575274-35575274 on Build GRCh38
rs866743376 Chr.17: 35575549-35575549 on Build GRCh38
rs754471266 Chr.17: 35575585-35575585 on Build GRCh38
rs111424374 Chr.17: 35575146-35575146 on Build GRCh38
rs886052830 Chr.17: 35575447-35575447 on Build GRCh38
rs1055712098 Chr.17: 35575517-35575517 on Build GRCh38
rs540347013 Chr.17: 35575336-35575336 on Build GRCh38
rs1045289989 Chr.17: 35575487-35575487 on Build GRCh38
rs975005955 Chr.17: 35575354-35575354 on Build GRCh38
rs539591621 Chr.17: 35575445-35575445 on Build GRCh38
rs886052829 Chr.17: 35575349-35575349 on Build GRCh38
rs555997949 Chr.17: 35575494-35575494 on Build GRCh38
rs944878975 Chr.17: 35575139-35575139 on Build GRCh38
rs961888073 Chr.17: 35575569-35575569 on Build GRCh38
rs987184149 Chr.17: 35575426-35575426 on Build GRCh38
rs912165834 Chr.17: 35575385-35575385 on Build GRCh38
rs573245836 Chr.17: 35575427-35575427 on Build GRCh38
rs935556612 Chr.17: 35575158-35575158 on Build GRCh38
rs890587764 Chr.17: 35575137-35575137 on Build GRCh38
rs549033406 Chr.17: 35575260-35575260 on Build GRCh38
rs920232689 Chr.17: 35575321-35575321 on Build GRCh38
rs959892912 Chr.17: 35575362-35575362 on Build GRCh38
rs886052828 Chr.17: 35575308-35575308 on Build GRCh38
rs192370696 Chr.17: 35575273-35575273 on Build GRCh38
rs939380551 Chr.17: 35575562-35575562 on Build GRCh38
rs200648389 Chr.17: 35575265-35575266 on Build GRCh38
rs915501223 Chr.17: 35575467-35575467 on Build GRCh38
rs959890795 Chr.17: 35575260-35575260 on Build GRCh38

View Full Product Details