Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35575511-35576020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
UniGene
Hs.591190
Gene Chromosome Location
Chr.17: 35574795-35578637 on Build GRCh38
Transcripts
NM_000286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725328 c.885C>T p.P295P Chr.17: 35575977-35575977 on Build GRCh38
6872224 c.861C>A p.H287Q Chr.17: 35576001-35576001 on Build GRCh38
4973502 c.980G>A p.R327H Chr.17: 35575882-35575882 on Build GRCh38
7089761 c.1069C>A p.P357T Chr.17: 35575793-35575793 on Build GRCh38
705498 c.1001G>T p.R334M Chr.17: 35575861-35575861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61752111 Chr.17: 35575975-35575975 on Build GRCh38
rs767070181 Chr.17: 35575978-35575978 on Build GRCh38
rs751033919 Chr.17: 35575743-35575743 on Build GRCh38
rs761445143 Chr.17: 35575890-35575890 on Build GRCh38
rs398123301 Chr.17: 35575973-35575974 on Build GRCh38
rs756694664 Chr.17: 35575744-35575744 on Build GRCh38
rs763619631 Chr.17: 35575738-35575738 on Build GRCh38
rs61752112 Chr.17: 35575913-35575913 on Build GRCh38
rs912131686 Chr.17: 35575653-35575653 on Build GRCh38
rs748308886 Chr.17: 35575811-35575811 on Build GRCh38
rs761590834 Chr.17: 35575976-35575976 on Build GRCh38
rs572785120 Chr.17: 35575727-35575727 on Build GRCh38
rs780946763 Chr.17: 35575747-35575747 on Build GRCh38
rs780595864 Chr.17: 35575930-35575930 on Build GRCh38
rs867245161 Chr.17: 35575951-35575952 on Build GRCh38
rs76127905 Chr.17: 35575928-35575928 on Build GRCh38
rs28936697 Chr.17: 35575903-35575903 on Build GRCh38
rs772194164 Chr.17: 35575950-35575950 on Build GRCh38
rs756569847 Chr.17: 35575765-35575765 on Build GRCh38
rs751429784 Chr.17: 35575883-35575883 on Build GRCh38
rs750097322 Chr.17: 35575916-35575916 on Build GRCh38
rs555645908 Chr.17: 35575684-35575684 on Build GRCh38
rs760739894 Chr.17: 35575853-35575853 on Build GRCh38
rs766933745 Chr.17: 35575909-35575909 on Build GRCh38
rs758276967 Chr.17: 35575733-35575733 on Build GRCh38
rs966726269 Chr.17: 35575726-35575726 on Build GRCh38
rs941358133 Chr.17: 35575884-35575884 on Build GRCh38
rs768427035 Chr.17: 35575972-35575972 on Build GRCh38
rs570189667 Chr.17: 35575554-35575554 on Build GRCh38
rs145574816 Chr.17: 35575959-35575959 on Build GRCh38
rs749859287 Chr.17: 35575990-35575990 on Build GRCh38
rs1037590 Chr.17: 35575616-35575616 on Build GRCh38
rs928085463 Chr.17: 35575802-35575802 on Build GRCh38
rs748595918 Chr.17: 35575947-35575947 on Build GRCh38
rs762912502 Chr.17: 35575865-35575865 on Build GRCh38
rs371277434 Chr.17: 35575956-35575956 on Build GRCh38
rs779617735 Chr.17: 35575940-35575940 on Build GRCh38
rs899106737 Chr.17: 35575663-35575663 on Build GRCh38
rs200789791 Chr.17: 35575815-35575815 on Build GRCh38
rs772028045 Chr.17: 35575841-35575841 on Build GRCh38
rs749650201 Chr.17: 35575898-35575901 on Build GRCh38
rs866743376 Chr.17: 35575549-35575549 on Build GRCh38
rs754471266 Chr.17: 35575585-35575585 on Build GRCh38
rs752689493 Chr.17: 35575746-35575746 on Build GRCh38
rs988425243 Chr.17: 35575729-35575729 on Build GRCh38
rs915485976 Chr.17: 35575754-35575754 on Build GRCh38
rs777654572 Chr.17: 35575955-35575955 on Build GRCh38
rs1047155507 Chr.17: 35575848-35575848 on Build GRCh38
rs755371958 Chr.17: 35575939-35575939 on Build GRCh38
rs373860467 Chr.17: 35575957-35575957 on Build GRCh38
rs61752109 Chr.17: 35575986-35575987 on Build GRCh38
rs780501878 Chr.17: 35575782-35575782 on Build GRCh38
rs755776824 Chr.17: 35575929-35575929 on Build GRCh38
rs373666248 Chr.17: 35575931-35575931 on Build GRCh38
rs532390390 Chr.17: 35575681-35575681 on Build GRCh38
rs762789314 Chr.17: 35575969-35575969 on Build GRCh38
rs954551964 Chr.17: 35575735-35575735 on Build GRCh38
rs776806092 Chr.17: 35575963-35575963 on Build GRCh38
rs986241167 Chr.17: 35575629-35575629 on Build GRCh38
rs961888073 Chr.17: 35575569-35575569 on Build GRCh38
rs398123302 Chr.17: 35575968-35575968 on Build GRCh38
rs752715446 Chr.17: 35575665-35575665 on Build GRCh38
rs557834416 Chr.17: 35575882-35575882 on Build GRCh38
rs28936697 Chr.17: 35575903-35575903 on Build GRCh38
rs774415243 Chr.17: 35575973-35575973 on Build GRCh38
rs981025397 Chr.17: 35575800-35575800 on Build GRCh38
rs771406750 Chr.17: 35575945-35575946 on Build GRCh38
rs769244620 Chr.17: 35575786-35575786 on Build GRCh38
rs1005986145 Chr.17: 35575608-35575608 on Build GRCh38
rs138720627 Chr.17: 35575995-35575995 on Build GRCh38
rs200283718 Chr.17: 35575860-35575860 on Build GRCh38
rs996017117 Chr.17: 35575673-35575673 on Build GRCh38
rs764657253 Chr.17: 35575874-35575875 on Build GRCh38
rs779530147 Chr.17: 35575794-35575794 on Build GRCh38
rs939380551 Chr.17: 35575562-35575562 on Build GRCh38
rs749833303 Chr.17: 35575784-35575784 on Build GRCh38
rs267608184 Chr.17: 35575813-35575815 on Build GRCh38
rs887088674 Chr.17: 35575593-35575593 on Build GRCh38

View Full Product Details