Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74916775-74917269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

SNP Detail

SNP ID Chromosome Location
rs943700161 Chr.17: 74916948-74916948 on Build GRCh38
rs938072206 Chr.17: 74917130-74917130 on Build GRCh38
rs146007618 Chr.17: 74917241-74917241 on Build GRCh38
rs3859210 Chr.17: 74916882-74916882 on Build GRCh38
rs189974618 Chr.17: 74916920-74916920 on Build GRCh38
rs749663203 Chr.17: 74917062-74917062 on Build GRCh38
rs1036907013 Chr.17: 74916921-74916921 on Build GRCh38
rs550587833 Chr.17: 74916967-74916967 on Build GRCh38
rs965006428 Chr.17: 74917047-74917047 on Build GRCh38
rs137943342 Chr.17: 74916979-74916979 on Build GRCh38
rs374828527 Chr.17: 74917006-74917007 on Build GRCh38
rs1001015730 Chr.17: 74917007-74917007 on Build GRCh38
rs912268146 Chr.17: 74917038-74917038 on Build GRCh38
rs112293979 Chr.17: 74917133-74917133 on Build GRCh38
rs959397720 Chr.17: 74916891-74916892 on Build GRCh38
rs977400332 Chr.17: 74917242-74917242 on Build GRCh38
rs746904393 Chr.17: 74916893-74916894 on Build GRCh38
rs902737106 Chr.17: 74917246-74917246 on Build GRCh38
rs945411833 Chr.17: 74916912-74916913 on Build GRCh38
rs535659863 Chr.17: 74916905-74916906 on Build GRCh38
rs995066862 Chr.17: 74917173-74917173 on Build GRCh38
rs761480955 Chr.17: 74916970-74916971 on Build GRCh38
rs143429562 Chr.17: 74916851-74916851 on Build GRCh38
rs142437952 Chr.17: 74917147-74917147 on Build GRCh38
rs888271506 Chr.17: 74916965-74916965 on Build GRCh38
rs888219256 Chr.17: 74917050-74917050 on Build GRCh38
rs911356647 Chr.17: 74917151-74917151 on Build GRCh38
rs113583471 Chr.17: 74917020-74917020 on Build GRCh38
rs568192417 Chr.17: 74917091-74917091 on Build GRCh38
rs1041317824 Chr.17: 74917028-74917028 on Build GRCh38
rs992590497 Chr.17: 74916903-74916904 on Build GRCh38
rs536064705 Chr.17: 74917021-74917021 on Build GRCh38
rs768758380 Chr.17: 74916940-74916941 on Build GRCh38
rs796824729 Chr.17: 74916892-74916893 on Build GRCh38
rs559327953 Chr.17: 74916943-74916943 on Build GRCh38
rs909296684 Chr.17: 74917092-74917092 on Build GRCh38
rs1034251000 Chr.17: 74916970-74916970 on Build GRCh38
rs966694229 Chr.17: 74917072-74917072 on Build GRCh38
rs1041139202 Chr.17: 74916951-74916952 on Build GRCh38
rs966866405 Chr.17: 74916802-74916802 on Build GRCh38
rs958191366 Chr.17: 74917002-74917002 on Build GRCh38
rs539014541 Chr.17: 74917199-74917199 on Build GRCh38
rs116670727 Chr.17: 74916975-74916975 on Build GRCh38
rs979955665 Chr.17: 74916883-74916890 on Build GRCh38
rs574552627 Chr.17: 74916800-74916800 on Build GRCh38
rs912475260 Chr.17: 74916883-74916883 on Build GRCh38
rs779459863 Chr.17: 74917186-74917186 on Build GRCh38
rs577242816 Chr.17: 74916865-74916865 on Build GRCh38
rs925404358 Chr.17: 74916916-74916925 on Build GRCh38
rs780234900 Chr.17: 74917055-74917055 on Build GRCh38
rs553241289 Chr.17: 74917222-74917222 on Build GRCh38
rs986057551 Chr.17: 74917005-74917005 on Build GRCh38
rs1032519138 Chr.17: 74917016-74917016 on Build GRCh38
rs756392131 Chr.17: 74917025-74917025 on Build GRCh38
rs918217926 Chr.17: 74917029-74917032 on Build GRCh38
rs115905441 Chr.17: 74917226-74917226 on Build GRCh38
rs936833612 Chr.17: 74916921-74916934 on Build GRCh38
rs549572410 Chr.17: 74917023-74917023 on Build GRCh38
rs896921870 Chr.17: 74916842-74916842 on Build GRCh38
rs762950394 Chr.17: 74916903-74916906 on Build GRCh38
rs1014550746 Chr.17: 74917059-74917059 on Build GRCh38
rs1049636049 Chr.17: 74916955-74916955 on Build GRCh38
rs1032411547 Chr.17: 74917113-74917113 on Build GRCh38
rs932326414 Chr.17: 74916857-74916857 on Build GRCh38
rs690566 Chr.17: 74916980-74916980 on Build GRCh38
rs1042678430 Chr.17: 74917230-74917230 on Build GRCh38
rs896524267 Chr.17: 74917136-74917136 on Build GRCh38
rs1048129198 Chr.17: 74917046-74917046 on Build GRCh38
rs952550315 Chr.17: 74916855-74916855 on Build GRCh38
rs918486002 Chr.17: 74917066-74917066 on Build GRCh38
rs114353953 Chr.17: 74916961-74916961 on Build GRCh38
rs751686069 Chr.17: 74916960-74916960 on Build GRCh38
rs886053385 Chr.17: 74916912-74916913 on Build GRCh38
rs537126229 Chr.17: 74917108-74917108 on Build GRCh38
rs541618266 Chr.17: 74916958-74916958 on Build GRCh38
rs978192363 Chr.17: 74916912-74916915 on Build GRCh38
rs931167517 Chr.17: 74917079-74917079 on Build GRCh38
rs368387485 Chr.17: 74916922-74916923 on Build GRCh38
rs993949629 Chr.17: 74916844-74916844 on Build GRCh38
rs561862648 Chr.17: 74916959-74916959 on Build GRCh38
rs182142592 Chr.17: 74916954-74916954 on Build GRCh38
rs911504003 Chr.17: 74916941-74916941 on Build GRCh38
rs111922548 Chr.17: 74916849-74916849 on Build GRCh38
rs760396641 Chr.17: 74917044-74917047 on Build GRCh38
rs895666575 Chr.17: 74917058-74917058 on Build GRCh38

View Full Product Details