Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021002.2 | NP_066282.1 | 1 - 1 | 352 | 88 |
GenBank mRNA | BC069471.1 | - | 1 - 1 | 352 | 88 |
BC096697.1 | - | 1 - 1 | 352 | 88 | |
BC096710.1 | - | 1 - 1 | 352 | 88 | |
BC096730.1 | - | 1 - 1 | 352 | 88 | |
BC098357.1 | - | 1 - 1 | 352 | 88 |