Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000208.3 | NP_000199.2 | 14 - 15 | 3256 | 137 |
NM_001079817.2 | NP_001073285.1 | 13 - 14 | 3220 | 137 | |
GenBank mRNA | AB208861.1 | - | 6 - 7 | 1198 | 137 |
BC117172.1 | - | 13 - 14 | 2842 | 137 | |
M10051.1 | - | 14 - 15 | 2983 | 137 | |
X02160.1 | - | 13 - 14 | 2857 | 137 |