Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000267.3 | NP_000258.1 | 29 - 30 | 4356 | 67 |
NM_001042492.2 | NP_001035957.1 | 29 - 30 | 4356 | 67 | |
XM_005257983.2 | XP_005258040.1 | 29 - 30 | 4306 | 67 | |
XM_005257984.2 | XP_005258041.1 | 29 - 30 | 4306 | 67 | |
XM_006721922.2 | XP_006721985.1 | 30 - 31 | 4336 | 67 | |
XM_006721924.2 | XP_006721987.1 | 30 - 31 | 4336 | 67 | |
XM_006721925.2 | XP_006721988.1 | 30 - 31 | 4336 | 67 | |
XM_006721926.3 | XP_006721989.1 | 30 - 31 | 4336 | 67 | |
XM_011524852.2 | XP_011523154.1 | 30 - 31 | 4333 | 67 | |
XM_011524857.2 | XP_011523159.1 | 30 - 31 | 4336 | 67 | |
XM_017024689.1 | XP_016880178.1 | 30 - 31 | 4336 | 67 | |
XM_017024690.1 | XP_016880179.1 | 30 - 31 | 4336 | 67 | |
GenBank mRNA | AB209336.1 | - | 12 - 13 | 2509 | 67 |
BC144643.1 | - | 29 - 30 | 4075 | 67 | |
BX537850.1 | - | 27 - 28 | 3748 | 67 | |
M38106.1 | - | 21 - 22 | 2971 | 67 | |
M38107.1 | - | 21 - 22 | 2971 | 67 | |
M60915.1 | - | 3 - 4 | 472 | 67 | |
M61213.1 | - | 5 - 6 | 706 | 67 | |
M82814.1 | - | 29 - 30 | 4184 | 67 | |
M89914.1 | - | 29 - 30 | 4170 | 67 |