Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000484.3 | NP_000475.1 | 10 - 11 | 1496 | 92 |
NM_001136016.3 | NP_001129488.1 | 9 - 10 | 1420 | 92 | |
NM_001136129.2 | NP_001129601.1 | 7 - 8 | 1103 | 92 | |
NM_001136130.2 | NP_001129602.1 | 9 - 10 | 1328 | 92 | |
NM_001136131.2 | NP_001129603.1 | 8 - 9 | 1158 | 92 | |
NM_001204301.1 | NP_001191230.1 | 10 - 11 | 1496 | 92 | |
NM_001204302.1 | NP_001191231.1 | 9 - 10 | 1439 | 92 | |
NM_001204303.1 | NP_001191232.1 | 8 - 9 | 1271 | 92 | |
NM_201413.2 | NP_958816.1 | 9 - 10 | 1439 | 92 | |
NM_201414.2 | NP_958817.1 | 8 - 9 | 1271 | 92 | |
GenBank mRNA | A02759.1 | - | 8 - 9 | 1217 | 92 |
AF282245.1 | - | 7 - 8 | 1030 | 92 | |
AK294534.1 | - | 9 - 10 | 1275 | 92 | |
AK295621.1 | - | 9 - 10 | 1381 | 92 | |
AK296229.1 | - | 8 - 9 | 1158 | 92 | |
AK297229.1 | - | 6 - 7 | 634 | 92 | |
AK297412.1 | - | 7 - 8 | 1000 | 92 | |
AK298861.1 | - | 7 - 8 | 1173 | 92 | |
AK312326.1 | - | 10 - 11 | 1361 | 92 | |
BC065529.1 | - | 9 - 10 | 1370 | 92 | |
M16765.1 | - | 3 - 4 | 231 | 92 | |
M28373.1 | - | 7 - 8 | 1002 | 92 | |
X06989.1 | - | 9 - 10 | 1363 | 92 | |
Y00264.1 | - | 8 - 9 | 1218 | 92 |