Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000924.3 | NP_000915.1 | 4 - 5 | 796 | 84 |
NM_001165975.2 | NP_001159447.1 | 3 - 4 | 484 | 84 | |
NM_001288768.1 | NP_001275697.1 | 4 - 5 | 858 | 84 | |
NM_001288769.1 | NP_001275698.1 | 3 - 4 | 416 | 84 | |
NM_001315534.1 | NP_001302463.1 | 3 - 4 | 387 | 84 | |
XM_011538456.1 | XP_011536758.1 | 5 - 6 | 746 | 84 | |
XM_017019432.1 | XP_016874921.1 | 3 - 4 | 315 | 84 | |
XM_017019433.1 | XP_016874922.1 | 3 - 4 | 1003 | 84 | |
GenBank mRNA | AB209200.1 | - | 4 - 5 | 565 | 84 |
AJ401609.1 | - | 3 - 4 | 343 | 84 | |
AK296422.1 | - | 3 - 4 | 416 | 84 | |
AK302931.1 | - | 3 - 4 | 471 | 84 | |
BC032226.1 | - | 4 - 5 | 569 | 84 | |
BP342896.1 | - | 3 - 4 | 463 | 84 | |
BX538092.1 | - | 3 - 4 | 387 | 84 | |
DC392024.1 | - | 3 - 4 | 471 | 84 | |
EU668337.1 | - | 4 - 5 | 565 | 84 | |
U56976.1 | - | 3 - 4 | 437 | 84 | |
U86078.1 | - | 4 - 5 | 516 | 84 |