Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001843.3 | NP_001834.2 | 20 - 21 | 2747 | 135 |
NM_175038.2 | NP_778203.1 | 18 - 19 | 2563 | 135 | |
XM_005268651.2 | XP_005268708.1 | 20 - 21 | 3110 | 135 | |
XM_006719241.2 | XP_006719304.1 | 20 - 21 | 2734 | 135 | |
XM_011537926.2 | XP_011536228.1 | 20 - 21 | 2699 | 135 | |
XM_011537927.2 | XP_011536229.1 | 20 - 21 | 2756 | 135 | |
XM_017018821.1 | XP_016874310.1 | 20 - 21 | 2750 | 135 | |
XM_017018822.1 | XP_016874311.1 | 20 - 21 | 2699 | 135 | |
XM_017018823.1 | XP_016874312.1 | 20 - 21 | 3110 | 135 | |
XM_017018824.1 | XP_016874313.1 | 20 - 21 | 2734 | 135 | |
GenBank mRNA | U07819.1 | - | 19 - 20 | 2520 | 135 |
U07820.1 | - | 18 - 19 | 2568 | 135 | |
Z21488.1 | - | 20 - 21 | 2641 | 135 |