Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001843.3 | NP_001834.2 | 22 - 23 | 3049 | 95 |
NM_175038.2 | NP_778203.1 | 20 - 21 | 2865 | 95 | |
XM_005268651.2 | XP_005268708.1 | 22 - 23 | 3412 | 95 | |
XM_006719241.2 | XP_006719304.1 | 22 - 23 | 3036 | 95 | |
XM_011537926.2 | XP_011536228.1 | 22 - 23 | 3001 | 95 | |
XM_011537927.2 | XP_011536229.1 | 22 - 23 | 3058 | 95 | |
XM_017018821.1 | XP_016874310.1 | 22 - 23 | 3052 | 95 | |
XM_017018822.1 | XP_016874311.1 | 22 - 23 | 3001 | 95 | |
XM_017018823.1 | XP_016874312.1 | 22 - 23 | 3412 | 95 | |
XM_017018824.1 | XP_016874313.1 | 22 - 23 | 3036 | 95 | |
GenBank mRNA | U07819.1 | - | 21 - 22 | 2822 | 95 |
U07820.1 | - | 20 - 21 | 2870 | 95 | |
Z21488.1 | - | 22 - 23 | 2943 | 95 |