Assay Design



Gene Transcripts

Gene Symbol
HTT
Entrez Gene ID
3064
Gene Name
huntingtin
Gene Aliases
HD, IT15
Chromosome Location
Chr.4: 3074510 - 3243960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002111.8 NP_002102.4 48 - 49 6779 62
GenBank mRNA AB016794.1 - 48 - 49 6779 62
AB209506.1 - 34 - 35 7756 62
L12392.1 - 48 - 49 6949 62

View Full Product Details