Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135651.2 | NP_001129123.1 | 10 - 11 | 1110 | 104 |
NM_002759.3 | NP_002750.1 | 10 - 11 | 1344 | 104 | |
XM_011532987.2 | XP_011531289.1 | 10 - 11 | 1044 | 104 | |
XM_017004503.1 | XP_016859992.1 | 10 - 11 | 1040 | 104 | |
GenBank mRNA | AK290655.1 | - | 10 - 11 | 1030 | 104 |
AK313818.1 | - | 9 - 10 | 867 | 104 | |
BC007769.2 | - | 9 - 10 | 886 | 104 | |
BC022314.1 | - | 10 - 11 | 998 | 104 | |
BC040851.1 | - | 10 - 11 | 1110 | 104 | |
BC057805.1 | - | 10 - 11 | 1015 | 104 | |
BC093676.1 | - | 8 - 9 | 815 | 104 | |
BC101475.1 | - | 8 - 9 | 815 | 104 | |
BC143228.1 | - | 8 - 9 | 814 | 104 | |
BC143230.1 | - | 8 - 9 | 815 | 104 | |
M35663.1 | - | 9 - 10 | 973 | 104 | |
M85294.1 | - | 8 - 9 | 817 | 104 |