Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004260.3 | NP_004251.3 | 2 - 3 | 164 | 78 |
XM_011517384.2 | XP_011515686.1 | 2 - 3 | 164 | 78 | |
XM_017013991.1 | XP_016869480.1 | 2 - 3 | 169 | 78 | |
XM_017013992.1 | XP_016869481.1 | 2 - 3 | 168 | 78 | |
XM_017013993.1 | XP_016869482.1 | 2 - 3 | 168 | 78 | |
XM_017013994.1 | XP_016869483.1 | 2 - 3 | 170 | 78 | |
XM_017013995.1 | XP_016869484.1 | 2 - 3 | 166 | 78 | |
XM_017013996.1 | XP_016869485.1 | 2 - 3 | 167 | 78 | |
XM_017013997.1 | XP_016869486.1 | 2 - 3 | 169 | 78 | |
XM_017013998.1 | XP_016869487.1 | 2 - 3 | 166 | 78 | |
XM_017013999.1 | XP_016869488.1 | 2 - 3 | 168 | 78 | |
GenBank mRNA | AB006532.1 | - | 2 - 3 | 78 | |
CN286057.1 | - | 2 - 3 | 165 | 78 | |
CN286060.1 | - | 2 - 3 | 156 | 78 |