Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004260.3 | NP_004251.3 | 8 - 9 | 1523 | 74 |
XM_011517384.2 | XP_011515686.1 | 8 - 9 | 1523 | 74 | |
XM_017013991.1 | XP_016869480.1 | 8 - 9 | 1528 | 74 | |
XM_017013992.1 | XP_016869481.1 | 8 - 9 | 1527 | 74 | |
XM_017013993.1 | XP_016869482.1 | 8 - 9 | 1527 | 74 | |
XM_017013994.1 | XP_016869483.1 | 9 - 10 | 1433 | 74 | |
XM_017013995.1 | XP_016869484.1 | 8 - 9 | 1525 | 74 | |
XM_017013996.1 | XP_016869485.1 | 8 - 9 | 1526 | 74 | |
XM_017013997.1 | XP_016869486.1 | 8 - 9 | 1528 | 74 | |
XM_017013998.1 | XP_016869487.1 | 8 - 9 | 1525 | 74 | |
XM_017013999.1 | XP_016869488.1 | 8 - 9 | 1527 | 74 | |
XM_017014000.1 | XP_016869489.1 | 4 - 5 | 439 | 74 | |
XM_017014001.1 | XP_016869490.1 | 6 - 7 | 1619 | 74 | |
GenBank mRNA | AB006532.1 | - | 8 - 9 | 1481 | 74 |
BC011602.2 | - | 1 - 2 | 82 | 74 | |
KJ147518.1 | - | 4 - 5 | 1028 | 74 |