Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000689.4 | NP_000680.2 | 10 - 11 | 1517 | 118 |
GenBank mRNA | AB209821.1 | - | 10 - 11 | 1244 | 118 |
AF003341.1 | - | 10 - 11 | 1200 | 118 | |
AK000118.1 | - | 10 - 11 | 1244 | 118 | |
AK026641.1 | - | 10 - 11 | 1244 | 118 | |
AK293176.1 | - | 10 - 11 | 1210 | 118 | |
AY390731.1 | - | 10 - 11 | 1200 | 118 | |
BC001505.2 | - | 10 - 11 | 1225 | 118 | |
BT006921.1 | - | 10 - 11 | 1200 | 118 | |
EU668318.1 | - | 10 - 11 | 1244 | 118 | |
EU794589.1 | - | 10 - 11 | 1200 | 118 | |
FJ224286.1 | - | 10 - 11 | 1253 | 118 | |
K03000.1 | - | 6 - 7 | 717 | 118 | |
M26761.1 | - | 6 - 7 | 717 | 118 |