Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000701.7 | NP_000692.2 | 13 - 14 | 2173 | 63 |
NM_001160233.1 | NP_001153705.1 | 13 - 14 | 2197 | 63 | |
NM_001160234.1 | NP_001153706.1 | 13 - 14 | 2006 | 63 | |
XM_017001360.1 | XP_016856849.1 | 14 - 15 | 5392 | 63 | |
XM_017001361.1 | XP_016856850.1 | 14 - 15 | 4734 | 63 | |
GenBank mRNA | AK293689.1 | - | 5 - 6 | 583 | 63 |
AK295095.1 | - | 13 - 14 | 2196 | 63 | |
AK296362.1 | - | 13 - 14 | 2006 | 63 | |
AK296384.1 | - | 13 - 14 | 2132 | 63 | |
AK308418.1 | - | 13 - 14 | 1938 | 63 | |
AK314777.1 | - | 13 - 14 | 2018 | 63 | |
BC003077.2 | - | 13 - 14 | 2086 | 63 | |
BC050359.1 | - | 13 - 14 | 2084 | 63 | |
J03007.1 | - | 8 - 9 | 1243 | 63 | |
U16798.1 | - | 12 - 13 | 1834 | 63 |