Assay Design



Gene Transcripts

Gene Symbol
C5AR1
Entrez Gene ID
728
Gene Name
complement component 5a receptor 1
Gene Aliases
C5A, C5AR, C5R1, CD88
Chromosome Location
Chr.19: 47309847 - 47322070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001736.3 NP_001727.1 2 - 2 1189 75
XM_005259190.4 XP_005259247.1 2 - 2 1162 75
GenBank mRNA BC008982.1 - 2 - 2 1217 75
M62505.1 - 2 - 2 1164 75

View Full Product Details