Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000207.2 | NP_000198.1 | 2 - 3 | 245 | 146 |
NM_001185097.1 | NP_001172026.1 | 2 - 3 | 271 | 146 | |
NM_001185098.1 | NP_001172027.1 | 1 - 2 | 424 | 146 | |
NM_001291897.1 | NP_001278826.1 | 2 - 3 | 305 | 146 | |
GenBank mRNA | AY899304.1 | - | 2 - 3 | 241 | 146 |
BC005255.1 | - | 2 - 3 | 245 | 146 | |
BM510347.1 | - | 2 - 3 | 266 | 146 | |
BM510748.1 | - | 2 - 3 | 249 | 146 | |
BP322143.1 | - | 1 - 2 | 424 | 146 | |
BT006808.1 | - | 1 - 2 | 186 | 146 | |
JF909299.1 | - | 1 - 2 | 139 | 146 | |
JQ951950.1 | - | 2 - 3 | 230 | 146 | |
X70508.1 | - | 2 - 3 | 230 | 146 |