Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000893.3 | NP_000884.1 | 3 - 4 | 626 | 129 |
NM_001102416.2 | NP_001095886.1 | 3 - 4 | 626 | 129 | |
NM_001166451.1 | NP_001159923.1 | 3 - 4 | 626 | 129 | |
GenBank mRNA | AI133186.1 | - | 3 - 4 | 536 | 129 |
AK223589.1 | - | 3 - 4 | 575 | 129 | |
AK290839.1 | - | 3 - 4 | 525 | 129 | |
AK298440.1 | - | 4 - 5 | 540 | 129 | |
AK303768.1 | - | 3 - 4 | 606 | 129 | |
AK315230.1 | - | 3 - 4 | 578 | 129 | |
BC060039.1 | - | 3 - 4 | 529 | 129 | |
CD014130.1 | - | 3 - 4 | 588 | 129 | |
K02566.1 | - | 3 - 4 | 441 | 129 |