Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030006.1 | NP_001025177.1 | 21 - 22 | 2973 | 94 |
NM_001282.2 | NP_001273.1 | 20 - 21 | 2931 | 94 | |
XM_005257937.3 | XP_005257994.1 | 21 - 22 | 3038 | 94 | |
XM_005257938.2 | XP_005257995.1 | 21 - 22 | 2939 | 94 | |
XM_011524448.1 | XP_011522750.1 | 22 - 23 | 3433 | 94 | |
XM_011524449.2 | XP_011522751.1 | 22 - 23 | 3134 | 94 | |
XM_011524450.1 | XP_011522752.1 | 22 - 23 | 3035 | 94 | |
XM_011524451.1 | XP_011522753.1 | 21 - 22 | 2962 | 94 | |
XM_011524452.1 | XP_011522754.1 | 23 - 24 | 3202 | 94 | |
XM_011524453.1 | XP_011522755.1 | 23 - 24 | 3120 | 94 | |
XM_011524454.1 | XP_011522756.1 | 23 - 24 | 3136 | 94 | |
XM_011524455.2 | XP_011522757.1 | 21 - 22 | 2982 | 94 | |
XM_017024284.1 | XP_016879773.1 | 20 - 21 | 2996 | 94 | |
XM_017024285.1 | XP_016879774.1 | 22 - 23 | 3106 | 94 | |
XM_017024286.1 | XP_016879775.1 | 21 - 22 | 3029 | 94 | |
GenBank mRNA | AK289528.1 | - | 21 - 22 | 2914 | 94 |
AK292531.1 | - | 21 - 22 | 2851 | 94 | |
AK295667.1 | - | 10 - 11 | 1123 | 94 | |
AK301522.1 | - | 21 - 22 | 2808 | 94 | |
AK304126.1 | - | 17 - 18 | 2328 | 94 | |
AY341427.1 | - | 21 - 22 | 2966 | 94 | |
BC006201.2 | - | 21 - 22 | 2880 | 94 | |
BC012150.1 | - | 12 - 13 | 1601 | 94 | |
CR749392.1 | - | 21 - 22 | 2956 | 94 | |
M34175.1 | - | 20 - 21 | 2919 | 94 |