Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002373.5 | NP_002364.5 | 2 - 3 | 172 | 126 |
XM_017022189.1 | XP_016877678.1 | 3 - 4 | 462 | 126 | |
GenBank mRNA | AK291052.1 | - | 2 - 3 | 144 | 126 |
BC055312.1 | - | 2 - 3 | 141 | 126 | |
BC094702.1 | - | 2 - 3 | 131 | 126 | |
BP309709.1 | - | 2 - 3 | 172 | 126 |