Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003019.4 | NP_003010.4 | 6 - 7 | 712 | 59 |
XM_011540087.1 | XP_011538389.1 | 6 - 7 | 691 | 59 | |
XM_011540088.2 | XP_011538390.1 | 5 - 6 | 663 | 59 | |
GenBank mRNA | BC022318.1 | - | 6 - 7 | 713 | 59 |
BM985387.1 | - | 2 - 3 | 59 | ||
CR541948.1 | - | 5 - 6 | 669 | 59 | |
X65018.1 | - | 6 - 7 | 840 | 59 |