Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004418.3 | NP_004409.1 | 2 - 3 | 594 | 96 |
XM_017003546.1 | XP_016859035.1 | 1 - 2 | 706 | 96 | |
GenBank mRNA | BC007771.2 | - | 2 - 3 | 570 | 96 |
L11329.1 | - | 2 - 3 | 593 | 96 |