Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004559.4 | NP_004550.2 | 7 - 8 | 1336 | 104 |
NR_132737.1 | 6 - 7 | 1154 | 104 | ||
GenBank mRNA | BC002411.2 | - | 7 - 8 | 1119 | 104 |
BC010430.1 | - | 7 - 8 | 1118 | 104 | |
BC013838.1 | - | 6 - 7 | 1154 | 104 | |
BC015208.1 | - | 7 - 8 | 1068 | 104 | |
BC020164.1 | - | 1 - 1 | 577 | 104 | |
BC038384.1 | - | 7 - 8 | 1125 | 104 | |
BC065571.1 | - | 7 - 8 | 1143 | 104 | |
BC071708.1 | - | 7 - 8 | 1128 | 104 | |
BC090038.1 | - | 7 - 8 | 1128 | 104 | |
BC098435.1 | - | 7 - 8 | 1134 | 104 | |
BC106045.1 | - | 7 - 8 | 1130 | 104 | |
J03827.1 | - | 7 - 8 | 1129 | 104 | |
L28809.1 | - | 7 - 8 | 1086 | 104 | |
M24070.1 | - | 7 - 8 | 1125 | 104 | |
M83234.1 | - | 1 - 1 | 1227 | 104 | |
M85234.1 | - | 1 - 1 | 1233 | 104 |