Assay Design



Gene Transcripts

Gene Symbol
TPSD1
Entrez Gene ID
23430
Gene Name
tryptase delta 1
Gene Aliases
MCP7-LIKE, MCP7L1, MMCP-7L
Chromosome Location
Chr.16: 1256272 - 1258493 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012217.2 NP_036349.1 4 - 5 692 108
GenBank mRNA AF206664.1 - 3 - 4 529 108
BC069143.1 - 4 - 5 683 108

View Full Product Details