Assay Design



Gene Transcripts

Gene Symbol
FAM173A
Entrez Gene ID
65990
Gene Name
family with sequence similarity 173 member A
Gene Aliases
C16orf24
Chromosome Location
Chr.16: 720581 - 722590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023933.2 NP_076422.1 4 - 5 588 68
GenBank mRNA BC001181.1 - 4 - 5 572 68
BC002624.2 - 4 - 5 540 68

View Full Product Details