Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031947.3 | NP_114153.1 | 1 - 1 | 1343 | 61 |
GenBank mRNA | AF332005.1 | - | 1 - 1 | 1145 | 61 |
AF378119.1 | - | 1 - 1 | 1325 | 61 | |
AY208943.1 | - | 1 - 1 | 1253 | 61 |